Skip to Content
Merck
All Photos(5)

Key Documents

HPA028897

Sigma-Aldrich

Anti-SMAD7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MADH7, Anti-MADH8, Anti-SMAD family member 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EVKRLCCCESYGKINPELVCCNPHHLSRLCELESPPPPYSRYPMDFLKPTADCPDAVPSSAETGGTNYLAPGGLSDSQLLLEPGDRSHWCVVAYWEEKTRVGRLYCVQEPSLDIFYDLPQGNG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD7(4092)

Looking for similar products? Visit Product Comparison Guide

General description

SMAD7 (mothers against decapentaplegic homolog 7) is a Tbx1 (T-box 1) interacting gene. It is an inhibitor of the TGFβ (transforming growth factor β) /BMP (bone morphogenetic proteins) pathway. It is located on chromosome 18q21.1.

Immunogen

SMAD family member 7 recombinant protein epitope signature tag (PrEST)

Application

Anti-SMAD7 has been used in immunohistochemistry.

Biochem/physiol Actions

SMAD7 (mothers against decapentaplegic homolog 7) is a modulator of TGFβ(transforming growth factor β) signaling in immune cells that are associated to ulcerative colitis and inflammatory bowel syndrome. It plays a major role in the etiology of CRC (colorectal cancer). It even functions as a mediator of the negative feedback loop for both the TGFβ and BMP (bone morphogenetic proteins) signaling pathways. It is essential for the remodelling of pharyngeal artery and the enlargement of great vessel. In mouse, homozygous removal of Smad7 causes primarily fourth-related arch artery defects. Silencing of Smad7 in RCD (refractory coeliac disease) biopsy samples can decrease the expression of interleukin-6 and tumour necrosis factor-α. Polymorphism of this gene is associated with colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86713

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vjera Mihaljević et al.
Biomedicines, 10(11) (2022-11-27)
Aims: Chronic diabetes complications, including diabetic nephropathy (DN), frequently result in end-stage renal failure. This study investigated empagliflozin (SGLT2i) effects on collagen synthesis, oxidative stress, cell survival, and protein expression in an LLC-PK1 model of DN. Methods: Combinations of high
High Smad7 sustains inflammatory cytokine response in refractory coeliac disease
Sedda S, et al.
Immunology, 150(3), 356-363 (2017)
Tbx1 genetically interacts with the transforming growth factor-?/bone morphogenetic protein inhibitor Smad7 during great vessel remodeling
Papangeli I and Scambler PJ
Circulation Research, 112(1), 90-102 (2013)
Intronic polymorphisms of the SMAD7 gene in association with colorectal cancer
Damavand B, et al.
Asian Pacific Journal of Cancer Prevention, 16(1), 41-44 (2015)
Multiple functional risk variants in a SMAD7 enhancer implicate a colorectal cancer risk haplotype
Fortini BK, et al.
PLoS ONE, 9(11) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service