Skip to Content
Merck
All Photos(6)

Key Documents

HPA023374

Sigma-Aldrich

Anti-PCM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-PCM-1, Anti-Pericentriolar material 1 protein, Anti-hPCM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TIYSEVATLISQNESRPHFLIELFHELQLLNTDYLRQRALYALQDIVSRHISESHEKGENVKSVNSGTWIASNSELTPSESLATTDDETFEKNFE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCM1(5108)

General description

The gene PCM1 (pericentriolar material 1) is mapped to human chromosome 8p22. The encoded protein localizes to cytoplasmic granules known as ‘centriolar satellites′, which are present around the centrosome. It has a molar mass of 228kDa protein.

Immunogen

Pericentriolar material 1 protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-PCM1 antibody has been used in cardiac myocytes nuclei (CM) staining and immunoblotting.

Biochem/physiol Actions

The gene PCM1 (pericentriolar material 1) encodes a large centrosomal protein with multiple coiled-coil domains that forms a complex with disrupted-in-schizophrenia 1 (DISC1) and Bardet-Biedl syndrome 4 protein (BBS4). This binding helps in synergistic targeting of PCM1 and other cargo proteins to the centrosome. It functions as a scaffold and facilitates the targeting of several proteins to the centrosome in a dynein motor-dependent manner and helps in regulating microtubular dynamics. The centriolar satellites that contain PCM1 protein participate in microtubule- and dynactin-dependent recruitment of proteins to the centrosome and mediate the assembly of centrosomal proteins and microtubule organization. Defects in this gene are associated with reduction in gray matter volume and susceptibility to schizophrenia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76188

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single cardiomyocyte nuclear transcriptomes reveal a lincRNA-regulated de-differentiation and cell cycle stress-response in vivo.
See K, et al.
Nature Communications, 8(1), 225-225 (2017)
DNA methylation signatures follow preformed chromatin compartments in cardiac myocytes.
Nothjunge S, et al.
Nature Communications, 8(1), 1667-1667 (2017)
A method for the acute and rapid degradation of endogenous proteins.
Clift D, et al.
Cell, 171(7), 1692-1706 (2017)
Kelvin See et al.
Nature communications, 8(1), 225-225 (2017-08-10)
Cardiac regeneration may revolutionize treatment for heart failure but endogenous progenitor-derived cardiomyocytes in the adult mammalian heart are few and pre-existing adult cardiomyocytes divide only at very low rates. Although candidate genes that control cardiomyocyte cell cycle re-entry have been
David K Kranzhöfer et al.
PloS one, 11(11), e0166575-e0166575 (2016-11-17)
Cardiomyocytes undergo major changes in DNA methylation during maturation and transition to a non-proliferative state after birth. 5'-hydroxylation of methylated cytosines (5hmC) is not only involved in DNA loss of CpG methylation but is also thought to be an epigenetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service