Skip to Content
Merck
All Photos(10)

Key Documents

HPA023300

Sigma-Aldrich

Anti-CTH antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Cystathionine gamma-lyase, Anti-Gamma-cystathionase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

TGCTGMVTFYIKGTLQHAEIFLKNLKLFTLAESLGGFESLAELPAIMTHASVLKNDRDVLGISDTLIRLSVGLEDEEDLLEDLDQALKAAHP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTH(1491)

General description

The gene CTH (cystathionine γ-lyase) is mapped to human chromosome 1p31. It is a member of the γ-family of pyridoxal phosphate (PLP)-dependent enzymes.

Immunogen

cystathionine gamma-lyase

Application

Anti-CTH antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

CTH (cystathionine γ-lyase) is a pyridoxal phosphate (PLP)-dependent enzyme which participates in the transsulfuration pathway. It is responsible for the generation of endogenous hydrogen sulfide (H2S) via desulfhydration of cysteine. In mouse model, absence of CTH causes hypertension and decreases endothelium-associated vasorelaxation. CTH-mediated H2S production protects against ischemic heart diseases.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75481

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P-N Teng et al.
British journal of cancer, 110(1), 123-132 (2013-11-02)
The majority of patients diagnosed with advanced epithelial ovarian carcinoma (EOC) relapse with resistant disease, and there are no biomarkers that possess clinical utility to identify or monitor these patients. This study aimed to identify secreted proteins ('secretome') collected from
Yaqi Shen et al.
Antioxidants & redox signaling, 22(3), 224-240 (2014-09-10)
Myocardial infarction (MI) is a leading cause of death globally. MicroRNAs (miRNAs) have been identified as a novel class of MI injury regulators. Hydrogen sulfide (H2S) is a gaseous signaling molecule that regulates cardiovascular function. The purpose of this study
Xing-Huang Gao et al.
Molecular & cellular proteomics : MCP, 19(5), 852-870 (2020-03-07)
The redox-based modifications of cysteine residues in proteins regulate their function in many biological processes. The gas molecule H2S has been shown to persulfidate redox sensitive cysteine residues resulting in an H2S-modified proteome known as the sulfhydrome. Tandem Mass Tags
Juan Carlos Souto et al.
American journal of human genetics, 76(6), 925-933 (2005-04-26)
Homocysteine (Hcy) plasma level is an independent risk marker for venous thrombosis, myocardial infarction, stroke, congestive heart failure, osteoporotic fractures, and Alzheimer disease. Hcy levels are determined by the interaction of genetic and environmental factors. The genetic basis is still
Alicia Madurga et al.
American journal of physiology. Lung cellular and molecular physiology, 309(7), L710-L724 (2015-08-02)
The gasotransmitter hydrogen sulfide (H2S) is emerging as a mediator of lung physiology and disease. Recent studies revealed that H2S administration limited perturbations to lung structure in experimental animal models of bronchopulmonary dysplasia (BPD), partially restoring alveolarization, limiting pulmonary hypertension

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service