Skip to Content
Merck
All Photos(7)

Documents

HPA019869

Sigma-Aldrich

Anti-GSTP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-GST class-pi, Anti-GSTP1-1, Anti-Glutathione S-transferase P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

TLGLYGKDQQEAALVDMVNDGVEDLRCKYISLIYTNYEAGKDDYVKALPGQLKPFETLLS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTP1(2950)

General description

GSTP1 (Glutathione S-transferase pi 1) is a novel cyclin dependent kinase-5 regulatory protein encoding a detoxifying enzyme. It is widely expressed in normal tissues.

Immunogen

Glutathione S-transferase P recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GSTP1 (Glutathione S-transferase pi 1) plays an important role in Cdk signaling pathway by protecting cells from DNA adduct formation. It prevent neurodegeneration, by blocking the activity of Cdk5 via p25/p35 translocation. It has been predicted that GSTP1 level may control Cdk5 signaling, and eliminate oxidative stress. It can also couple with a wide number of exogenous and endogenous hydrophobic electrophiles. Overexpression of enzyme activity has been reported in several types of cancers, such as blood, head, and neck, lung, colorectal, esophagus and breast cancers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74589

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Jie Xiao et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 11(20), e2306059-e2306059 (2024-03-26)
Tumor-initiating cells (TICs) resilience and an immunosuppressive microenvironment are aggressive oncogenic phenotypes that contribute to unsatisfactory long-term outcomes in lung adenocarcinoma (LUAD) patients. The molecular mechanisms mediating the interaction between TICs and immune tolerance have not been elucidated. The role
Si-Qi Wang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 10(7), e2205262-e2205262 (2023-01-30)
Glutathione S-transferase pi (GSTP1), a phase II detoxification enzyme, is known to be overexpressed and mediates chemotherapeutic resistance in lung cancer. However, whether GSTP1 supports cancer stem cells (CSCs) and the underlying mechanisms in lung adenocarcinoma (LUAD) remain largely unknown. This study unveiled
Yusuke Yamamoto et al.
Oncology reports, 30(4), 1687-1694 (2013-07-12)
Glutathione S-transferases (GSTs) have been reported to be activated in several types of cancers, including esophageal squamous cell carcinoma (ESCC). The aim of the present study was to investigate whether GSTP1 protein expression is a useful predictor of the clinical
Yue Jia et al.
Experimental and molecular pathology, 108, 156-163 (2019-04-06)
As the fifth most common cancer and the second leading cause of cancer related deaths worldwide, hepatocellular carcinoma (HCC) causes up to one million deaths annually. Alcoholic steatohepatitis (ASH) and non-alcoholic steatohepatitis (NASH) are becoming the two major risk factors
D Pellacani et al.
Cell death and differentiation, 21(5), 761-773 (2014-01-28)
Prostate cancer (CaP) is mostly composed of luminal-like differentiated cells, but contains a small subpopulation of basal cells (including stem-like cells), which can proliferate and differentiate into luminal-like cells. In cancers, CpG island hypermethylation has been associated with gene downregulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service