Skip to Content
Merck
All Photos(6)

Key Documents

HPA019208

Sigma-Aldrich

Anti-BTN2A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BT2.1, Anti-BTF1, Anti-BTN2.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ILSGEKEFERETREIALKELEKERVQKEEELQVKEKLQEELRWRRTFLHAVDVVLD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BTN2A1(11120)

General description

The gene BTN2A1 (butyrophilin subfamily 2 member A1) is mapped to human chromosome 6p22.1. It is ubiquitously expressed. BTN2A1 is an MHC (major histocompatibility complex)-encoded cell surface glycoprotein.

Immunogen

Butyrophilin subfamily 2 member A1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BTN2A1 (butyrophilin subfamily 2 member A1) is suggested to have roles in lipid, fatty acid, and sterol metabolism. Polymorphisms in BTN2A1 are linked with dyslipidemia, myocardial infarction, type 2 diabetes mellitus, chronic kidney disease and hypertension.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72842

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideki Horibe et al.
Molecular medicine reports, 9(3), 808-812 (2014-01-24)
We have previously shown that the C→T polymorphism (rs6929846) of the butyrophilin, subfamily 2, member A1 gene (BTN2A1) was significantly associated with myocardial infarction. Considering that dyslipidemia is a significant risk factor for coronary heart disease, it was hypothesized that
M Hiramatsu et al.
Diabetic medicine : a journal of the British Diabetic Association, 28(11), 1381-1387 (2011-06-16)
We previously showed that the C→T polymorphism (rs6929846) of BTN2A1 was significantly associated with myocardial infarction in Japanese individuals by a genome-wide association study. Given that diabetes mellitus is an important risk factor for myocardial infarction, the association of rs6929846
Georg Malcherek et al.
Journal of immunology (Baltimore, Md. : 1950), 179(6), 3804-3811 (2007-09-06)
The MHC-encoded butyrophilin, BTN2A1, is a cell surface glycoprotein related to the extended family of B7 costimulatory molecules. BTN2A1 mRNA was expressed in most human tissues, but protein expression was significantly lower in leukocytes. An Ig-fusion protein of BTN2A1 bound
Yoshiji Yamada et al.
Atherosclerosis, 215(1), 145-152 (2011-01-08)
We have performed a genome-wide association study (GWAS) to identify genetic variants that confer susceptibility to myocardial infarction (MI) in Japanese and Korean populations. A total of 17,447 Japanese or Korean individuals from four independent subject panels was examined. Japanese
Hideki Horibe et al.
American journal of hypertension, 24(8), 924-929 (2011-04-29)
We previously showed that the C→T polymorphism (rs6929846) in butyrophilin, subfamily 2, member A1 gene (BTN2A1) was associated with myocardial infarction in Japanese individuals. Given that hypertension is a major risk factor for myocardial infarction, the association of rs6929846 of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service