Skip to Content
Merck
All Photos(5)

Key Documents

HPA019032

Sigma-Aldrich

Anti-TET1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CXXC-type zinc finger protein 6, Anti-Leukemia-associated protein with a CXXC domain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VPPNPIATFNAPSKWPEPQSTVSYGLAVQGAIQILPLGSGHTPQSSSNSEKNSLPPVMAISNVENEKQVHISFLPANTQGFPLAPERGLFHASLGIAQLSQAGPSKSDRGSSQVSVTSTVHVVNTTVVTMPVPMVSTSSSSYTT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TET1(80312)

General description

TET1 is a protein that belongs to the TET family and contains a zinc-binding CXXC domain. TET1 functions as a 5-methylcytosine (5mC) hydroxylase and converts 5-methylcytosine to 5-hydroxymethylcytosine which is crucial for active DNA demethylation in the adult brain. Anti-TET1 antibody can be used in western blotting. Rabbit anti-TET1 antibody reacts specifically with TET1 protein.

Immunogen

Protein TET1 (Ten-eleven translocation 1 gene protein) (CXXC-type zinc finger protein 6) (Leukemia-associated protein with a CXXC domain)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74805

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elham Barazeghi et al.
Clinical epigenetics, 8, 31-31 (2016-03-15)
Primary hyperparathyroidism is characterized by enlarged parathyroid glands due to an adenoma (80-85 %) or multiglandular disease (~15 %) causing hypersecretion of parathyroid hormone (PTH) and generally hypercalcemia. Parathyroid cancer is rare (<1-5 %). The epigenetic mark 5-hydroxymethylcytosine (5hmC) is reduced in various
Fu Zhao et al.
Frontiers in oncology, 11, 603686-603686 (2021-03-16)
Medulloblastoma, as the most common malignant brain tumor in children, exhibits highly dysregulated DNA methylation. The novel epigenetic marker-5-hydroxymethylcytosine (5hmC) plays essential role in gene regulation during brain development and in brain tumors. However, the biological and clinical implications of
Sarah Teuber-Hanselmann et al.
International journal of molecular sciences, 22(8) (2021-05-01)
Quantifying O6-methylguanine-DNA methyltransferase (MGMT) promoter methylation plays an essential role in assessing the potential efficacy of alkylating agents in the chemotherapy of malignant gliomas. MGMT promoter methylation is considered to be a characteristic of subgroups of certain malignancies but has
Shuang-Ling Wu et al.
Genomics, proteomics & bioinformatics, 19(1), 64-79 (2021-03-16)
Although DNA 5-hydroxymethylcytosine (5hmC) is recognized as an important epigenetic mark in cancer, its precise role in lymph node metastasis remains elusive. In this study, we investigated how 5hmC associates with lymph node metastasis in breast cancer. Accompanying with high
Ryoichi Ono et al.
Cancer research, 62(14), 4075-4080 (2002-07-19)
There are a limited number of reports of acute myeloid leukemia (AML) with t(10;11)(q22;q23). We showed that the MLL gene on 11q23 was fused to the LCX (leukemia-associated protein with a CXXC domain) gene on 10q22 in a de novoadult

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service