Skip to Content
Merck
All Photos(6)

Documents

HPA017991

Sigma-Aldrich

Anti-FZD6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Frizzled-6 precursor, Anti-Fz-6, Anti-hFz6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

TSTGATANHGTSAVAITSHDYLGQETLTEIQTSPETSMREVKADGASTPRLREQDCGEPASPAASISRLSGEQVDGKGQAGSVSESARSEGRISPKSDITDTGLAQSNNLQVPSSSEPSSLKGSTSLLVHPVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD6(8323)

General description

Frizzled class receptor 6 (FZD6) is a 706 amino-acid, membrane-bound receptor. It possesses seven transmembrane domains, an N-terminal region with a cysteine-rich domain and two N-linked glycosylation sites. The gene encoding FZD6 is localized on chromosome 8q22.3.

Immunogen

frizzled class receptor 6

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Frizzled class receptor 6 (FZD6) acts a molecular switch between various Wnt pathways. FZD6 is needed for the genes of the non-canonical pathway to be expressed and the growth of the nail plate in humans. Mutations in the gene encoding it have been linked to nail dysplasia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72691

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Cantilena et al.
Oncotarget, 2(12), 976-983 (2012-01-18)
Wnt signalling is an important component of vertebrate development, required for specification of the neural crest. Ten Wnt receptors [Frizzled receptor 1-10 (Fzd1-10)] have been identified so far, some of which are expressed in the developing nervous system and the
G Naz et al.
The British journal of dermatology, 166(5), 1088-1094 (2012-01-04)
Isolated nail dysplasia is rare and has been reported in only a small number of families. To describe and characterize two Pakistani families with an autosomal-recessive inherited nail dysplasia. Genome-wide linkage analysis; mutation screening of candidate genes by Sanger sequencing;
M Tokuhara et al.
Biochemical and biophysical research communications, 243(2), 622-627 (1998-03-03)
The Frizzled genes encode receptors for WNTs, secreted glycoproteins implicated in development as well as in carcinogenesis. In this paper, we report molecular cloning of Hfz6, the human homologue of Mfz6. Nucleotide sequence analysis showed that the Hfz6 gene encodes
S I Raza et al.
The British journal of dermatology, 168(2), 422-425 (2012-08-07)
Inherited isolated nail anomaly manifesting with onychauxis and onycholysis is a rare condition, caused by mutations in the gene FZD6, encoding membrane-bound Wnt receptor protein. To search for sequence variants in the gene FZD6 in three individuals of a consanguineous
Zhenzhen Chen et al.
Oncogene, 37(23), 3098-3112 (2018-03-15)
Liver tumor-initiating cells (TICs), the drivers for liver tumorigenesis, accounts for liver tumor initiation, metastasis, drug resistance and relapse. Wnt/β-catenin signaling pathway emerges as a critical modulator in liver TIC self-renewal. However, the molecular mechanism of Wnt/β-catenin initiation in liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service