Skip to Content
Merck
All Photos(3)

Key Documents

HPA017917

Sigma-Aldrich

Anti-KRT1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-67 kDa cytokeratin, Anti-CK-1, Anti-Cytokeratin-1, Anti-Hair alpha protein, Anti-K1, Anti-Keratin, type II cytoskeletal 1, Anti-Keratin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

MSRQFSSRSGYRSGGGFSSGSAGIINYQRRTTSSSTRRSGGGGGRFSSCGGGGGSFGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT1(3848)

General description

Keratin 1, type II (KRT1) is a protein of the cytoplasmic intermediate filament expressed in keratinocytes. KRT1 has an amino-terminal domain, a non-helical carboxy terminal domain and two rod domains. The gene encoding this protein is localized on human chromosome 12.

Immunogen

Keratin, type II cytoskeletal 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-KRT1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-KRT1 antibody produced in rabbit has been used for cytofluorimetric analysis.

Biochem/physiol Actions

Keratin 1, type II (KRT1) is a marker of epithelial differentiation and mutation in the gene encoding it is associated with epidermolytic hyperkeratosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74604

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Chen et al.
Journal of immunology research, 2014, 492849-492849 (2014-07-23)
Early diagnosis and prognosis monitoring are very important for the survival of patients with bladder cancer. To identify candidate biomarkers of bladder cancer, we used a combination of techniques including 2-DE, co-IP, western blot, LC-MS/MS, and immunohistochemistry. Hsp74 was identified
Mild phenotype of epidermolytic hyperkeratosis mimicking ichthyosis bullosa of Siemens is related to specific mutation in 2B domain of KRT1.
Jae Yong Sung et al.
Journal of dermatological science, 70(3), 220-222 (2013-04-30)
Keith A Choate et al.
The Journal of clinical investigation, 125(4), 1703-1707 (2015-03-17)
Widespread reversion of genetic disease is rare; however, such events are particularly evident in some skin disorders in which normal clones develop on a background of affected skin. We previously demonstrated that mutations in keratin 10 (KRT10) cause ichthyosis with
Marilena Gallotta et al.
Infection and immunity, 82(7), 2890-2901 (2014-04-30)
Group A streptococcus (GAS) is a human pathogen causing a wide repertoire of mild and severe diseases for which no vaccine is yet available. We recently reported the identification of three protein antigens that in combination conferred wide protection against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service