Skip to Content
Merck
All Photos(6)

Documents

HPA017357

Sigma-Aldrich

Anti-NT5E antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CALJA, Anti-CD73, Anti-NT5, Anti-eN, Anti-eNT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

EFDNGVEGLIEPLLKEAKFPILSANIKAKGPLASQISGLYLPYKVLPVGDEVVGIVGYTSKETPFLSNPGTNLVFEDEITALQPEVDKLKTLNVNKIIALGHSGFEMDKLIAQK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NT5E(4907)

General description

NT5E (5′-nucleotidase, ecto, CD73) is a glycosyl-phosphatidylinositol-linked plasma membrane glycoprotein encoding a protein, ecto-5′-nucleotidase (CD73), which acts as the major enzymatic source of extracellular adenosine. It is expressed in different types of cells and in different tissues.

Immunogen

5′-nucleotidase precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-NT5E antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

NT5E (5′-nucleotidase, ecto, CD73) plays an important role in the stabilization of tissue homeostasis and controlling of several pathophysiologic responses related to immunity, inflammation, pain, ischemia, tissue fibrosis, and cancer. It is involved in the generation of adenosine from degradation of AMP (adenosine monophosphate) in the extracellular environment. CD73 facilitates the conversion of intracellular AMP to adenosine, which further regulates various processes by activating one of four subtypes of G-protein-coupled receptors. Mutation in NT5E causes an adult-onset condition characterized by joint pain, calcification of joints and arteries (CALJA).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72880

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Reinhardt et al.
Cancer research, 77(17), 4697-4709 (2017-06-28)
Evolution of tumor cell phenotypes promotes heterogeneity and therapy resistance. Here we found that induction of CD73, the enzyme that generates immunosuppressive adenosine, is linked to melanoma phenotype switching. Activating MAPK mutations and growth factors drove CD73 expression, which marked
Randy S Haun et al.
Cancer biology & therapy, 16(10), 1557-1565 (2015-07-16)
To develop new diagnostic and therapeutic tools to specifically target pancreatic tumors, it is necessary to identify cell-surface proteins that may serve as potential tumor-specific targets. In this study we used an azido-labeled bioorthogonal chemical reporter to metabolically label N-linked
Natasha T Snider et al.
Molecular biology of the cell, 25(25), 4024-4033 (2014-10-10)
Ecto-5'-nucleotidase (CD73), encoded by NT5E, is the major enzymatic source of extracellular adenosine. CD73 controls numerous pathophysiological responses and is a potential disease target, but its regulation is poorly understood. We examined NT5E regulation by alternative splicing. Genomic database analysis
Ilaria Magagna et al.
Cancers, 13(23) (2021-12-11)
Cancer-associated fibroblasts (CAF) are heterogeneous with multiple functions in breast cancer. Recently, we identified a specific CAF subpopulation (referred to as CAF-S1), which promotes immunosuppression and immunotherapy resistance. Here, by studying a large collection of human samples, we highlight the
Michel Fausther et al.
PloS one, 9(6), e98568-e98568 (2014-06-03)
Ecto-5'-nucleotidase/CD73/NT5E, the product of the NT5E gene, is the dominant enzyme in the generation of adenosine from degradation of AMP in the extracellular environment. Nonsense (c.662C→A, p.S221X designated F1, c.1609dupA, p.V537fsX7 designated F3) and missense (c.1073G→A, p.C358Y designated F2) NT5E

Articles

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service