Skip to Content
Merck
All Photos(6)

Key Documents

HPA017353

Sigma-Aldrich

Anti-KCNJ5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CIR, Anti-GIRK4, Anti-KATP1, Anti-Kir3.4, Anti-LQT13

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VTPWDPKKIPKQARDYVPIATDRTRLLAEGKKPRQRYMEKSGKCNVHHGNVQETY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ5(3762)

Immunogen

G protein-activated inward rectifier potassium channel 4 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72858

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Jen Cheng et al.
The Journal of clinical endocrinology and metabolism, 100(1), E155-E163 (2014-10-28)
Aldosterone-producing adenoma (APA) has been linked to mutations in the KCNJ5 gene encoding the inward-rectifying potassium (K(+)) Kir3.4 channel. These mutations abolish the K(+) selectivity of Kir3.4 and, consequently, cause sodium (Na(+)) leak, depolarized membrane potential, and nonsuppressible aldosterone secretion.
Takashi Okamura et al.
Endocrine journal, 64(1), 39-47 (2016-09-30)
Somatic mutations in KCNJ5 gene have been identified in patients with adrenal aldosterone-producing adenomas (APAs). We previously reported that Japanese patients with APAs had distinct characteristics from patients in Western countries; i.e. they had a high frequency of KCNJ5 mutations
Elena A B Azizan et al.
The Journal of clinical endocrinology and metabolism, 97(5), E819-E829 (2012-03-24)
Aldosterone-producing adenomas (APA) are heterogeneous. The recent finding of somatic KCNJ5 mutations suggests a genetic explanation. The objectives of this study were the following: 1) to compare transcriptional profiles in APA and adjacent adrenal gland (AAG); 2) to test whether
Iris Hardege et al.
Molecular endocrinology (Baltimore, Md.), 29(10), 1522-1530 (2015-09-05)
Primary aldosteronism accounts for 5%-10% of hypertension and in a third of cases is caused by autonomous aldosterone production by adenomas (APA). Somatic mutations in the potassium channel encoded by KCNJ5 have been detected in surgically removed APAs. To better
Junhua Zhou et al.
Hypertension (Dallas, Tex. : 1979), 65(5), 1103-1110 (2015-03-18)
Common somatic mutations in CACNAID and ATP1A1 may define a subgroup of smaller, zona glomerulosa (ZG)-like aldosterone-producing adenomas. We have therefore sought signature ZG genes, which may provide insight into the frequency and pathogenesis of ZG-like aldosterone-producing adenomas. Twenty-one pairs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service