Skip to Content
Merck
All Photos(6)

Key Documents

HPA015876

Sigma-Aldrich

Anti-HSPB8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Alpha-crystallin C chain, Anti-E2-induced gene 1 protein, Anti-Heat shock protein beta-8, Anti-HspB8, Anti-Protein kinase H11, Anti-Small stress protein-like protein HSP22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
recombinant expression
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LLDDGFGMDPFPDDLTASWPDWALPRLSSAWPGTLRSGMVPRGPTATARFGVPAEGRTPPPFPGEPWKVCVNVHSFKPEELMVKTKDGYVEVSGKHEEKQQEGGIVSKNFTKKIQLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPB8(26353)

Immunogen

Heat shock protein beta-8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Heat shock protein β-8 is a protein encoded by the HSPB8 gene in humans. It is a small HSP expressed ubiquitously in various tissues that serves as a positive regulator in TGF-α-induced migration of ovarian cancer cells. This gene is associated with aggressiveness and prognosis of tumor cells, thereby protecting tumor cell from apoptosis in many malignancies. Mutation in this gene (i.e. missense mutation) may lead to distal hereditary motor neuropathy (dHMN) and axonal Charcot-Marie-Tooth disease (CMT2L).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73509

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sejad Al-Tahan et al.
Neurology. Genetics, 5(4), e349-e349 (2019-08-14)
We clinically and molecularly characterize a new family with autosomal dominant rimmed vacuolar myopathy (RVM) caused by mutations in the HSPB8 gene. We performed whole-exome and whole-genome sequencing in the family. Western blot and immunocytochemistry were used to analyze 3
Alice S Kwok et al.
Journal of neurochemistry, 119(6), 1155-1161 (2011-10-12)
HspB8, a small heat-shock protein implicated in autophagy, is mutated in patients with distal hereditary motor neuropathy type II (dHMNII). Autophagy is essential for maintaining protein homeostasis in the central nervous system, but its role has not been investigated in
Joy Irobi et al.
Neuromuscular disorders : NMD, 22(8), 699-711 (2012-05-19)
Missense mutations in the small heat shock protein HSPB8 cause distal hereditary motor neuropathy (dHMN) and axonal Charcot-Marie-Tooth disease (CMT2L). We previously demonstrated that, despite the ubiquitous expression of HSPB8, motor neurons appear to be predominantly affected by HSPB8 mutations.
Mariko Suzuki et al.
Archives of biochemistry and biophysics, 571, 40-49 (2015-03-04)
Accumulating evidence suggests that heat shock proteins (HSPs) are implicated in progression of cancer. HSP22 (HSPB8), a small HSP, is recognized to be ubiquitously expressed in various tissues. However, the expression and the role of HSP22 in ovarian cancer remain
Xiao-shan Li et al.
Journal of cancer research and clinical oncology, 140(8), 1305-1313 (2014-05-09)
The heat shock protein 22 (HSP22) is associated with tumor proliferation and protects tumor cell from apoptosis in many malignancies. However, the role of HSP22 in gastric cancer has not been thoroughly elucidated. The aim was to determine the relationship

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service