Skip to Content
Merck
All Photos(4)

Key Documents

HPA015652

Sigma-Aldrich

Anti-BNIP3L antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Adenovirus E1B19K-binding protein B5, Anti-BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, Anti-BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, Anti-NIP3-like protein X, Anti-NIP3L

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SNGNDNGNGKNGGLEHVPSSSSIHNGDMEKILLDAQHESGQSSSRGSSHCDSPSPQEDGQIMFDVEMHTSRDHSSQSEEEVVEGEKEVEALKKSADWVSDWSSRPENIPPKEFHFRHPKRSVSLSMRKSGAMK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BNIP3L(665)

Immunogen

BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like recombinant protein epitope signature tag (PrEST)

Application

Anti-BNIP3L antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like is a protein encoded by the BNIP3L gene in humans. It is a tumor suppressor gene located at chromosome 8p21. It functions in promoting apoptosis by interacting with antiapoptotic proteins (such as Bcl-2, Bcl-x(L), E1B19K, Nix) and as a tumor suppressor. It is involved in lung cancer development and occurrence, when down-regulated.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71725

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuntao Lu et al.
PloS one, 7(9), e44559-e44559 (2012-09-18)
Previous reports indicate that the NIX/BNIP3L gene acts as a pro-apoptotic factor by interacting with BCL2 and BCL-XL, playing an important role in hypoxia-dependent cell death and acting as a tumor suppressor. However, many studies also showed that NIX is
Wulin Aerbajinai et al.
Blood, 102(2), 712-717 (2003-03-29)
Transcriptional profiles of cultured primary human erythroid cells were examined to identify those genes involved in the control of erythroid growth during the terminal phase of maturation. Our in silico screening strategy indicated that a hypoxia-inducible proapoptotic member of the
M Matsushima et al.
Genes, chromosomes & cancer, 21(3), 230-235 (1998-04-02)
We have isolated a novel cDNA that encodes a product showing significant sequence homology (56% identity) to human NIP3, a protein thought to interact with adenovirus E1B19kD and human BCL2 proteins. This cDNA contains an open reading frame of 657
Ji-Li Sun et al.
Ai zheng = Aizheng = Chinese journal of cancer, 23(1), 8-14 (2004-01-15)
Bcl-2/E1B 19kDa interacting protein3-like (BNIP3L) gene is a tumor suppressor gene cloned from a human fetal liver cDNA library, which is located at 8p21, one of the high frequent regions of loss of heterozygosity (LOH) in lung carcinoma. BNIP3L protein
Wen Li et al.
The Journal of biological chemistry, 289(15), 10691-10701 (2014-02-28)
Mitophagy receptors mediate the selective recognition and targeting of damaged mitochondria by autophagosomes. The mechanism for the regulation of these receptors remains unknown. Here, we demonstrated that a novel hypoxia-responsive microRNA, microRNA-137 (miR-137), markedly inhibits mitochondrial degradation by autophagy without

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service