Skip to Content
Merck
All Photos(7)

Key Documents

HPA015076

Sigma-Aldrich

Anti-SGMS2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Phosphatidylcholine:ceramide cholinephosphotransferase 2, Anti-Sphingomyelin synthase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

IIETAKLEEHLENQPSDPTNTYARPAEPVEEENKNGNGKPKSLSSGLRKGTKKYPDYIQIAMPTESR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGMS2(166929)

General description

SGMS2 (sphingomyelin synthase 2), also called SMS2 is one of the two isoforms of SMS enzyme, which is the last enzyme of the sphingomyelin (SM) synthesis pathway. SMS1, SMS2 and SMSr form the complete SMS family. It resides in the plasma membrane, and its cytoplasmic C-terminal is palmitoylated at its cysteine residues. It contains four motifs called D1, D2, D3 and D4, which are highly conserved in nature. It has six transmembrane α-helices, and D1 motif is located in the first exoplasmic loop and D2 in the third transmembrane α-helix.

Immunogen

Phosphatidylcholine:ceramide cholinephosphotransferase 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SGMS2 (sphingomyelin synthase 2) is the last enzyme in the de novo sphingomyelin (SM) synthesis pathway, which occurs in the plasma membrane. Studies in mice show that this enzyme promotes atherogenesis, and might have potential as a therapeutic target of atherosclerosis. It is also a part of the ceramide phosphoethanolamine biosynthesis. In various cancer cell types, it plays an essential role in cell growth. A compound with anti-tumor activity, called 2-hydroxyoleic acid (2OHOA) causes the activation of SGMS2, which leads to significant increase in the SM amount in plasma membrane. This activation is related to 2OHOA capability to induce cell cycle arrest and apoptosis in cancer cells. This protein also induces NF-κB pathway, which in turn has proatherogenic activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73281

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiruneh K Hailemariam et al.
Arteriosclerosis, thrombosis, and vascular biology, 28(8), 1519-1526 (2008-06-21)
NFkappaB has long been regarded as a proatherogenic factor, mainly because of its regulation of many of the proinflammatory genes linked to atherosclerosis. Metabolism of sphingomyelin (SM) has been suggested to affect NFkappaB activation, but the mechanism is largely unknown.
Motohiro Tani et al.
Biochemical and biophysical research communications, 381(3), 328-332 (2009-02-24)
Sphingomyelin synthase (SMS) is an enzyme that catalyzes the transfer of phosphocholine from phosphatidylcholine to ceramide for sphingomyelin synthesis. Here, we show that SMS2 is palmitoylated at cysteine residues via thioester bonds in the COOH-terminal cytoplasmic tail. [3H]palmitic acid labeling
Xiaogang Wang et al.
Lipids in health and disease, 10, 7-7 (2011-01-18)
Sphingomyelin synthase 2 (SMS2) contributes to de novo sphingomyelin (SM) biosynthesis. Its activity is related to SM levels in the plasma and the cell membrane. In this study, we investigated the possibility of a direct relationship between SMS and atherosclerosis.
Philipp Ternes et al.
Journal of lipid research, 50(11), 2270-2277 (2009-05-21)
Sphingolipids are vital components of eukaryotic membranes involved in the regulation of cell growth, death, intracellular trafficking, and the barrier function of the plasma membrane (PM). While sphingomyelin (SM) is the major sphingolipid in mammals, previous studies indicate that mammalian
Klazien Huitema et al.
The EMBO journal, 23(1), 33-44 (2003-12-20)
Sphingomyelin (SM) is a major component of animal plasma membranes. Its production involves the transfer of phosphocholine from phosphatidylcholine onto ceramide, yielding diacylglycerol as a side product. This reaction is catalysed by SM synthase, an enzyme whose biological potential can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service