Skip to Content
Merck
All Photos(1)

Key Documents

HPA013994

Sigma-Aldrich

Anti-NEBL antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Actin-binding Z-disk protein, Anti-Nebulette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

CTELISDIRYKEEFKKSKDKCTFVTDSPMLNHVKNIGAFISEAKYKGTIKADLSNSLYKRMPATIDSVFAGEVTQLQSEVAYKQKHDAAKGFSDYAHMKEPPEVKHAMEVNKHQSNISYRKDVQDTHTYSAELDRPD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEBL(10529)

General description

The gene NEBL (nebulette) encodes a nebulin like protein having a molar mass of 107kDa. It is predominantly expressed in the cardiac muscle. The protein is very similar to C-terminal region of human nebulin, an actin-binding skeletal muscle protein. They share a family of highly related C-terminal repeats, a serine-rich domain, and an SH3 domain. The gene is mapped to human chromosome 10p12. A short linker domain links the 22 nebulin-like repeats to a C-terminal Src homology 3 (SH3) domain. The repeat domain binds to actin filaments and the linker domain targets this association to Z-lines and dense bodies. The linker and SH3 domains are capable of differentiating between dense bodies and Z-lines. The alternative splice variant called LIM-nebulette is expressed in non-muscle cells. It interacts with Zyxin via the SH3 domain along with Lasp-1 and functions in the organization of focal adhesions.

Immunogen

Nebulette recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The protein, nebulette, encoded by the gene NEBL is found to be expressed aligned with the nascent myofibrils early during myofibrillogenesis. It may function in myofibril organization and function. It also interacts with I-Z-I complex of cardiac myofibrils and functions in the assembly of the Z-line.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72556

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiupeng Zhang et al.
Molecular carcinogenesis, 56(12), 2558-2565 (2017-07-02)
Lasp2, as well as Lasp1, is a member of the LIM-protein subfamily of the nebulin group characterized by the combined presence of LIM and SH3 domains. Lasp1 and Lasp2 are highly conserved in their LIM, nebulin-like, and SH3 domains but
C L Moncman et al.
Cell motility and the cytoskeleton, 32(3), 205-225 (1995-01-01)
A 107-kD protein has been identified in primary cultures of chicken embryonic cardiomyocytes by immunoprecipitations with certain anti-nebulin monoclonal antibodies (mAbs). These mAbs, prepared against a fragment of human skeletal muscle nebulin located near the carboxyl terminus, detect a 107-kD
Bo Li et al.
The Journal of biological chemistry, 279(19), 20401-20410 (2004-03-09)
Zyxin is a versatile component of focal adhesions in eukaryotic cells. Here we describe a novel binding partner of zyxin, which we have named LIM-nebulette. LIM-nebulette is an alternative splice variant of the sarcomeric protein nebulette, which, in contrast to
C L Moncman et al.
Cell motility and the cytoskeleton, 44(1), 1-22 (1999-09-02)
Nebulette, a 107 kDa protein associated with the I-Z-I complex of cardiac myofibrils, may play an important role in the assembly of the Z-line. Determination of the complete primary structure of 1011 residue human fetal nebulette reveals a four-domain layout
Carole L Moncman et al.
Experimental cell research, 273(2), 204-218 (2002-02-02)
To evaluate nebulette's role in cardiac myofibrils, cardiomyocytes expressing green fluorescent protein (GFP)-nebulette constructs were monitored for their ability to contract and myofilament protein distribution was analyzed. Cells expressing full-length GFP-nebulette appear unaffected and exhibit normal beating frequencies. Expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service