Skip to Content
Merck
All Photos(2)

Key Documents

HPA012887

Sigma-Aldrich

Anti-ATP7A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Copper pump 1, Anti-Copper-transporting ATPase 1, Anti-Menkes disease-associated protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

TETLGTCIDFQVVPGCGISCKVTNIEGLLHKNNWNIEDNNIKNASLVQIDASNEQSSTSSSMIIDAQISNALNAQQYKVLIGNREWMIRNGLVINNDVNDFMTEHERKGRTAVLVAVDDELC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP7A(538)

General description

Copper-transporting ATPase 1 (ATP7A) is a P-type ATPase localized in the trans-Golgi network (TGN). It has an amino-terminal region with six metal binding motifs, eight transmembrane domains and several other residues or motifs generally present in heavy metal-transporting ATPases. The gene encoding this protein is localized on chromosome Xq13.3.

Immunogen

Copper-transporting ATPase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Copper-transporting ATPase 1 (ATP7A) is mainly involved in delivering copper to copper-requiring enzymes. At times when cells are exposed to excessive copper, it is rapidly relocalized from the trans-Golgi network (TGN) to the plasma membrane and takes care of copper efflux. Thus, its functions include the transport of copper to cuproenzymes and proteins in the secretory pathway as well as excretion of excess copper. It constantly cycles between the TGN and plasma membrane. Mutations in the gene encoding ATP7A have been linked to Menkes disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74406

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Petris et al.
Human molecular genetics, 8(11), 2107-2115 (1999-09-15)
Menkes disease is an X-linked recessive copper deficiency disorder caused by mutations in the ATP7A ( MNK ) gene which encodes a copper transporting P-type ATPase (MNK). MNK is normally localized pre- dominantly in the trans -Golgi network (TGN); however
M Qi et al.
Human molecular genetics, 7(3), 465-469 (1998-04-18)
The ATP7A gene encodes a copper-transporting ATPase. Mutations in this gene result in two clinically distinct X-linked inherited disorders: Menkes disease and occipital horn syndrome (OHS). We identified a single exon skipping in the ATP7A transcript in cells from the
Z Tümer et al.
Advances in experimental medicine and biology, 448, 83-95 (1999-03-18)
Our knowledge about Menkes disease (MD) has expanded greatly since its description in 1962 as a new X-linked recessive neurodegenerative disorder of early infancy. Ten years later a defect in copper metabolism was established as the underlying biochemical deficiency. In
Sarah E M Stephenson et al.
The Journal of biological chemistry, 280(39), 33270-33279 (2005-07-30)
The homeostatic regulation of essential elements such as copper requires many proteins whose activities are often mediated and tightly coordinated through protein-protein interactions. This regulation ensures that cells receive enough copper without intracellular concentrations reaching toxic levels. To date, only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service