Skip to Content
Merck
All Photos(2)

Key Documents

HPA012567

Sigma-Aldrich

Anti-CYP26B1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cytochrome P450 26B1, Anti-P450 26A2, Anti-P450 retinoic acid-inactivating 2, Anti-P450RAI-2, Anti-Retinoic acid-metabolizing cytochrome

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SIPEEDLGHLFEVYQQFVDNVFSLPVDLPFSGYRRGIQARQILQKGLEKAIREKLQCTQGKDYLDALDLLIESSKEHGKEMTMQELKDGTLELIFAAYATTASASTSLIM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP26B1(56603)

Immunogen

Cytochrome P450 26B1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CYP26B1 (Cytochrome P450, family 26, subfamily B, polypeptide) is an inhibitor of all-trans-retinoic acid (RA). It is expressed in the cerebellum and pons of human adult brain. In addition to inactivation of RA, it also regulates RA metabolism. It is also involved in both level of patterning and in bone ossification during skeletal development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71472

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nelly Frydman et al.
Human reproduction (Oxford, England), 32(3), 631-642 (2017-01-12)
How can pre-meiotic germ cells persist in the human foetal ovary? Numerous oogonia escaping meiotic entry were retrieved throughout human ovarian development simultaneously with the expression of signalling pathways preventing meiosis, typically described in the rodent embryonic testis. The transition
Katarina Danzl et al.
Journal of molecular and cellular cardiology, 126, 105-117 (2018-11-26)
Physiologically, following myocardial infarction (MI), retinoid levels elevate locally in the infarcted area. Whereas therapeutic systemic application of retinoids was shown to reduce the progression of ventricular dilatation and the onset of heart failure, the role of acute physiologically increased
Anne Jørgensen et al.
The Journal of pathology, 229(4), 588-598 (2013-01-11)
Testicular germ cell tumours (TGCT) of young adults arise from the intratubular precursor, carcinoma in situ (CIS). CIS cells are thought to be developmentally arrested and transformed fetal germ cells that survive through childhood and gain invasive capacity after puberty.
J A White et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(12), 6403-6408 (2000-05-24)
Retinoids, particularly all-trans-retinoic acid (RA), are potent regulators of cell differentiation, cell proliferation, and apoptosis. The role of all-trans-RA during development and in the maintenance of adult tissues has been well established. The control of all-trans-RA levels in cells and
Kathrin Laue et al.
American journal of human genetics, 89(5), 595-606 (2011-10-25)
Excess exogenous retinoic acid (RA) has been well documented to have teratogenic effects in the limb and craniofacial skeleton. Malformations that have been observed in this context include craniosynostosis, a common developmental defect of the skull that occurs in 1

Articles

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service