Skip to Content
Merck
All Photos(6)

Key Documents

HPA010806

Sigma-Aldrich

Anti-B4GALT1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-β-1,4-GalTase 1 antibody produced in rabbit, Anti-β-1,4-Galactosyltransferase 1 antibody produced in rabbit, Anti-β4Gal-T1 antibody produced in rabbit, Anti-UDP-Gal:β-GlcNAc β-1,4-galactosyltransferase 1 antibody produced in rabbit, Anti-UDP-galactose:β-N-acetylglucosamine β-1,4-galactosyltransferase 1 antibody produced in rabbit, Anti-b4Gal-T1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

FRGMSISRPNAVVGRCRMIRHSRDKKNEPNPQRFDRIAHTKETMLSDGLNSLTYQVLDVQRYPLYTQITVDIGTPS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... B4GALT1(2683)

Looking for similar products? Visit Product Comparison Guide

General description

B4GALT1 (UDP-Gal:βGlcNAc β 1,4- galactosyltransferase, polypeptide 1) belongs to a family of type II transmembrane proteins called glycosyltransferases, which resides in Golgi apparatus. This family consists of seven members, from GALT1 to GALT7. B4GALT1 has two isoforms, having different cellular locations, because of differences in their cytoplasmic domains. The long isoform is localized to the cell surface, whereas the short isoform is found in the Golgi apparatus. This gene is located on human chromosome 9p13, and codes for a protein containing 398 amino acids. It is expressed in most tissues, excluding adult brain and fetal heart and brain.

Immunogen

β-1,4-Galactosyltransferase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

B4GALT1 (UDP-Gal:βGlcNAc β 1,4- galactosyltransferase, polypeptide 1) is responsible for the synthesis of galactose β-1,4-N-acetylglucosamine (Galβ1-4GlcNAc) groups on glycoproteins, on their N-linked sugar chains. This is performed by the short isoform of B4GALT1, which resides in Golgi bodies. The long isoform, localized to the cell surface, helps in cell adhesion, by interacting with N-acetylglucosamine containing oligosaccharides, which are found in the extracellular matrix. In patients with rheumatoid arthritis, it is involved in the inflammatory response in the synovial tissue. B4GALT1 plays an important role in the proliferation of MCF-7 breast cancer cells, by interacting with estrogen receptor. It is methylated and down-regulated in colorectal cancer patients, and hence, can act as a marker of invasive phenotype of colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71675

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huyang Xie et al.
BMC cancer, 18(1), 590-590 (2018-05-26)
The expression alterations of B4GALT1 have been noted in some types of cancer and they are related to cancer cell proliferation, invasiveness, metastasis, and drug resistance. We aimed to establish the expression of B4GALT1 in bladder cancer and its connection
Huyang Xie et al.
Oncotarget, 7(22), 32723-32730 (2016-04-20)
B4GALT1 is one of seven beta-1, 4-galactosyltransferase (B4GALT) genes, which has distinct functions in various malignances. Here, we evaluate the association of B4GALT1 expression with oncologic outcome in patients with non-metastatic clear cell renal cell carcinoma (ccRCC). A retrospective analysis
Maïlys Guillard et al.
The Journal of pediatrics, 159(6), 1041-1043 (2011-09-17)
The clinical phenotype of congenital disorders of glycosylation is heterogeneous, mostly including a severe neurological involvement and multisystem disease. We identified a novel patient with a galactosyltransferase deficiency with mild hepatopathy and coagulation anomalies, but normal psychomotor development. The tissue-specific
Maria Luana Poeta et al.
Genes, chromosomes & cancer, 51(12), 1133-1143 (2012-08-29)
Epigenetic alterations, such as CpG islands methylation and histone modifications, are recognized key characteristics of cancer. Glycogenes are a group of genes which epigenetic status was found to be changed in several tumors. In this study, we determined promoter methylation
Hee-Jung Choi et al.
Biochemical and biophysical research communications, 426(4), 620-625 (2012-09-18)
Beta 1,4-galactosyltransferase 1 (B4GALT1) synthesizes galactose β-1,4-N-acetylglucosamine (Galβ1-4GlcNAc) groups on N-linked sugar chains of glycoproteins, which play important roles in many biological events, including the proliferation and migration of cancer cells. A previous microarray study reported that this gene is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service