Skip to Content
Merck
All Photos(7)

Key Documents

HPA009084

Sigma-Aldrich

Anti-SIGLEC6 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD327 antigen, Anti-CD33 antigen-like 1, Anti-CDw327, Anti-OB-BP1, Anti-Obesity-binding protein 1, Anti-Sialic acid-binding Ig-like lectin 6 precursor, Anti-Siglec-6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

RVKTRRKKAAQPVQNTDDVNPVMVSGSRGHQHQFQTGIVSDHPAEAGPISEDEQELHYAVLHFHKVQPQEPKVTDTEYSEIKIHK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIGLEC6(946)

Looking for similar products? Visit Product Comparison Guide

General description

SIGLEC6 (sialic acid binding Ig like lectin 6) is a primate specific transmembrane receptor, which belongs to the Siglec or CD33 related protein family. In humans, this protein is exclusively expressed in placenta.

Immunogen

Sialic acid-binding Ig-like lectin 6 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SIGLEC6 (sialic acid binding Ig like lectin 6) is expressed in preterm preeclampsia (PE) pregnancy placenta, where it might contribute to or be a response to pathogenesis of PE. This protein is a binding-partner of sialyl-Tn glycans and leptin. The expression of this protein is linked with the onset, tempo and duration of human birth process. SIGLEC6, along with leptin, are overexpressed in gestational trophoblastic disease, where they promote proliferation, invasion and apoptosis. Interaction of SIGLEC6 with glycodelin-A leads to inhibition of extracellular signal-regulated kinase (ERK)/c-Jun signaling pathway, which results in the suppression of the invasiveness of trophoblast.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71738

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingxin Yu et al.
Frontiers in immunology, 9, 2138-2138 (2018-10-09)
Mast cells (MC) accumulate in colorectal cancer (CRC) and the relationship between MC density and cancer progression has been well recognized. MC can be either pro-tumor or anti-tumor players, depending on the local factors present in the tumor microenvironment. Upon
Jessica Nunes et al.
Nature communications, 15(1), 5180-5180 (2024-06-19)
Siglec-6 is a lectin receptor with restricted expression in the placenta, mast cells and memory B-cells. Although Siglec-6 is expressed in patients with chronic lymphocytic leukemia (CLL), its pathophysiological role has not been elucidated. We describe here a role for
Kevin K W Lam et al.
The Journal of biological chemistry, 286(43), 37118-37127 (2011-09-02)
During placentation, the cytotrophoblast differentiates into the villous cytotrophoblast and the extravillous cytotrophoblast. The latter invades the decidualized endometrium. Glycodelin-A (GdA) is abundantly synthesized by the decidua but not the trophoblast. Previous data indicate that GdA suppresses the invasion of
Kristen K Rumer et al.
Reproductive sciences (Thousand Oaks, Calif.), 20(6), 646-653 (2012-11-23)
Sialic acid immunoglobulin-like lectin (Siglec)-6 is a transmembrane receptor that binds sialyl-TN glycans and leptin. Among eutherian mammals, only human placentas express Siglec-6. Previous work has implicated Siglec-6 in preeclampsia (PE). Preeclampsia, a leading cause of maternal and perinatal morbidity
Kristen K Rumer et al.
Endocrine-related cancer, 19(6), 827-840 (2012-10-24)
Sialic acid immunoglobulin-like lectin (Siglec)-6 is a transmembrane receptor that binds leptin. Leptin is an obesity-associated peptide hormone overexpressed in gestational trophoblastic disease (GTD). GTD encompasses several placental abnormalities that range from benign to malignant. Among GTD, molar placentas are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service