Skip to Content
Merck
All Photos(1)

Key Documents

HPA008257

Sigma-Aldrich

Anti-TGFBR3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-βGlycan antibody produced in rabbit, Anti-TGF-β receptor type III precursor antibody produced in rabbit, Anti-TGFR-3 antibody produced in rabbit, Anti-Transforming growth factor-β receptor III antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GYSGMDVTLLDPTCKAKMNGTHFVLESPLNGCGTRPRWSALDGVVYYNSIVIQVPALGDSSGWPDGYEDLESGDNGFPGDMDEGDASLFTRPEIVVFNCSLQQVRNPSSFQEQPHGNITFNMELYNTDLF

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGFBR3(7049)

General description

The gene TGFBR3 (Transforming growth factor β receptor III) is mapped to human chromosme 1p32-p33.

Immunogen

TGF-β receptor type III precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Transforming growth factor β (TGF-β) interacts with TGF β receptor III (TGFBR3) which allows TGF-β to form a high affinity ternary comple with TGF-β receptor type II (TGFBRII). Thus, TGFBR3 plays an important role as a coreceptor to increase ligand binding to TGFBRII. It enhances TGF-β-mediated effects on the cells and eliminates the biological differences between TGF-β isoforms. TGFBR3 inhibits breast cancer cell invasion by shedding its ectodomain and producing soluble TGFBR3. Soluble TGFBR3 binds and sequesters TGF-β to decrease its signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70460

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F López-Casillas et al.
Cell, 73(7), 1435-1444 (1993-07-02)
Transforming growth factor beta (TGF beta) signals through a heteromeric protein kinase receptor that has a limited ability to bind ligand. This limitation is overcome by the action of betaglycan (TGF beta type III receptor), a separate TGF beta-binding membrane
Martin G Dalin et al.
Nature communications, 8(1), 1197-1197 (2017-11-01)
Myoepithelial carcinoma (MECA) is an aggressive salivary gland cancer with largely unknown genetic features. Here we comprehensively analyze molecular alterations in 40 MECAs using integrated genomic analyses. We identify a low mutational load, and high prevalence (70%) of oncogenic gene
Mei Dong et al.
The Journal of clinical investigation, 117(1), 206-217 (2006-12-13)
The TGF-beta signaling pathway has a complex role in regulating mammary carcinogenesis. Here we demonstrate that the type III TGF-beta receptor (TbetaRIII, or betaglycan), a ubiquitously expressed TGF-beta coreceptor, regulated breast cancer progression and metastasis. Most human breast cancers lost
Assignment of human transforming growth factor-beta type I and type III receptor genes (TGFBR1 and TGFBR3) to 9q33-q34 and 1p32-p33, respectively.
D W Johnson et al.
Genomics, 28(2), 356-357 (1995-07-20)
Anne M Kouri et al.
Kidney international reports, 8(11), 2368-2375 (2023-11-29)
Primary membranous nephropathy (PMN) is uncommon in children. Therefore, data on the clinical course of affected children are scarce. In recent years, several novel antigens have been implicated in the pathogenesis of PMN. However, the histopathologic characteristics of pediatric patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service