Skip to Content
Merck
All Photos(8)

Key Documents

HPA005750

Sigma-Aldrich

Anti-WASL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-N-WASP, Anti-Neural Wiskott-Aldrich syndrome protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DHQVPTTAGNKAALLDQIREGAQLKKVEQNSRPVSCSGRDALLDQIRQGIQLKSVADGQESTPPTPAPTSGIVGALMEVMQKRS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WASL(8976)

Immunogen

Neural Wiskott-Aldrich syndrome protein recombinant protein epitope signature tag (PrEST)

Application

Anti-WASL antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Neural Wiskott-Aldrich syndrome protein is a protein encoded by the WASL gene in humans. It is mapped on to chromosome 7q31.3. It is referred as NWASP and N-WASP. It is a major cytoskeletal regulator that stimulates Arp2/3-mediated actin nucleation. It regulates the cytoskeleton and junctional integrity at the epithelial zonula adherens. It is a junctional protein and depletion decreased junctional F-actin content and organization. It plays a role in assembly of kinetochores and attachment of microtubules to chromosomes, essential for accurate chromosome congression and segregation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70077

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamie A Whitelaw et al.
Cells, 9(7) (2020-07-11)
Cells migrating over 2D substrates are required to polymerise actin at the leading edge to form lamellipodia protrusions and nascent adhesions to anchor the protrusion to the substrate. The major actin nucleator in lamellipodia formation is the Arp2/3 complex, which
M Fukuoka et al.
Gene, 196(1-2), 43-48 (1997-10-10)
We recently identified Neural Wiskott-Aldrich Syndrome Protein (N-WASP) from bovine brain. An expression analysis using bovine cDNA revealed that N-WASP plays critical roles in the regulation of the cortical actin cytoskeleton. Here, we report the molecular cloning of N-WASP homologs
Eva M Kovacs et al.
Nature cell biology, 13(8), 934-943 (2011-07-26)
N-WASP is a major cytoskeletal regulator that stimulates Arp2/3-mediated actin nucleation. Here, we identify a nucleation-independent pathway by which N-WASP regulates the cytoskeleton and junctional integrity at the epithelial zonula adherens. N-WASP is a junctional protein whose depletion decreased junctional
Adrian Pătru et al.
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie, 61(4), 1259-1278 (2021-06-26)
Oral cancer remains an important global health issue and despite recent diagnostic and therapeutic advances, it continues to have an unfavorable prognostic and decreased survival. Although palatal tumors represent one of the rarest locations of oral squamous cell carcinomas (SCCs)
Sun Joo Park et al.
Molecules and cells, 31(6), 515-521 (2011-05-03)
The accurate distribution and segregation of replicated chromosomes through mitosis is crucial for cellular viability and development of organisms. Kinetochores are responsible for the proper congression and segregation of chromosomes. Here, we show that neural Wiskott-Aldrich syndrome protein (N-WASP) localizes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service