Skip to Content
Merck
All Photos(8)

Documents

HPA004807

Sigma-Aldrich

Anti-MEF2D antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myocyte-specific enhancer factor 2D antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VPVSNQSSLQFSNPSGSLVTPSLVTSSLTDPRLLSPQQPALQRNSVSPGLPQRPASAGAMLGGDLNSANGACPSPVGNGYVSARASPGLLPVANGNSLNKVIPAKSPPPPTHSTQLGAPSRKPDLRVITSQAG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MEF2D(4209)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Myocyte-specific enhancer factor 2D recombinant protein epitope signature tag (PrEST)

Application

Anti-MEF2D antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

MEF2D (myocyte enhancer factor 2D) is a member of MEF2 family. It is majorly involved in the muscle and neuronal differentiation as a transcriptional activator. It is also associated with cardiac morphogenesis, blood vessel formation, and growth factor responsiveness. It has been reported that the presence of MEF2D activates muscle specific luciferase constructs. As a consequence, it increases expression of the muscle-specific gene, myosin heavy chain, and a marker for skeletal muscle differentiation. It has also been concluded that MEF2D is upregulated in the p21 cells. In hepatocellular carcinoma, MEF2D regulates transcription of G2-M transition-retarding genes. Thus, it proves that MEF2D is a tumor-promoting gene in human hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86940

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nianxu Luan et al.
Oncology letters, 16(1), 1173-1179 (2018-07-03)
The microRNA (miR)-30 family has been reported to be aberrantly expressed in several types of cancer. However, its contributions to lung cancer remain to be fully elucidated. Myocyte enhancer factor 2D (MEF2D), an oncogene in liver cancer, has been shown
Meiling Zhang et al.
Molecular cancer, 12(1), 150-150 (2013-11-28)
Rhabdomyosarcoma (RMS) is a highly malignant pediatric cancer that is the most common form of soft tissue tumors in children. RMS cells have many features of skeletal muscle cells, yet do not differentiate. Thus, our studies have focused on the
Youguang Zhao et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 37(1), 601-610 (2015-08-04)
The prognosis of patients with malignant glioma is always quite poor, and this poor prognosis is probably due to our incomplete understanding of the molecular mechanisms underlying malignant glioma. It is known that myocyte enhancer factor-2D (MEF2D) plays an oncogenic
Leina Ma et al.
Cancer research, 74(5), 1452-1462 (2014-01-07)
The underlying molecular pathogenesis in hepatocellular carcinoma remains poorly understood. The transcription factor MEF2D promotes survival in various cell types and it seems to function as an oncogene in leukemia. However, its potential contributions to solid cancers have not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service