Skip to Content
Merck
All Photos(1)

Documents

HPA003180

Sigma-Aldrich

Anti-TNRC6B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Trinucleotide repeat-containing 6B protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QIPQFQLACQLLLQQQQQQQLLQNQRKISQAVRQQQEQQLARMVSALQQQQQQQQRQPGMKHSPSHPVGPKPHLDNMVPNALNVGLPDLQTKGPIPGYGSGFSSGGMDYGMVGGKEAGTESRFKQWTS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNRC6B(23112)

General description

The gene TNRC6B (trinucleotide repeat containing 6B) is mapped to human chromosome 22q13.1. The encoded protein contains high content of GW/WG repeats, one or more Q/P-rich motifs, and a C-terminal RNA recognition motif (RRM) domain. The RRM domain is essential for the protein to carry out translational repression.

Immunogen

Trinucleotide repeat-containing 6B protein recombinant protein epitope signature tag (PrEST)

Application

Anti-TNRC6B antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

TNRC6B (trinucleotide repeat containing 6B) gene encodes an RNA recognition motif (RRM)-containing protein that belongs to the GW182 family of proteins. It associates with argonaute (Ago) proteins involved in RNA silencing processes that are guided by both micro-RNAs (miRNAs) and short interfering RNAs (siRNAs). This association also represses translation and promotes degradation of mRNA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73563

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prestige Antibodies--monospecific antibodies designed for immunohistochemical analysis.
Sara Gunnerås et al.
BioTechniques, 44(6), 825-828 (2008-05-15)
Yoshiki Murakami et al.
BMC cancer, 13, 99-99 (2013-03-19)
Many studies support the hypothesis that specific microRNA (miRNA) expression in various human cancers including hepatocarcinogenesis is closely associated with diagnosis and prognosis. In hepatocellular carcinoma (HCC), malignancy level is related to the degree of histological differentiation. In order to
Todd L Edwards et al.
Human genetics, 132(12), 1361-1369 (2013-07-31)
Uterine fibroids (UFs) affect 77 % of women by menopause and account for $9.4 billion in yearly healthcare costs. We recently replicated findings from the first UF genome-wide association study (GWAS), conducted in the Japanese. Here we tested these GWAS-discovered
Gunter Meister et al.
Current biology : CB, 15(23), 2149-2155 (2005-11-18)
RNA silencing processes are guided by small RNAs known as siRNAs and microRNAs (miRNAs) . They reside in ribonucleoprotein complexes, which guide the cleavage of complementary mRNAs or affect stability and translation of partial complementary mRNAs . Argonaute (Ago) proteins
Joerg E Braun et al.
Molecular cell, 44(1), 120-133 (2011-10-11)
miRNAs are posttranscriptional regulators of gene expression that associate with Argonaute and GW182 proteins to repress translation and/or promote mRNA degradation. miRNA-mediated mRNA degradation is initiated by deadenylation, although it is not known whether deadenylases are recruited to the mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service