Skip to Content
Merck
All Photos(6)

Documents

HPA002858

Sigma-Aldrich

Anti-SYP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MRX96

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DMDVVNQLVAGGQFRVVKEPLGFVKVLQWAAPSVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYP(6855)

Immunogen

Synaptophysin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Synaptophysin (also known as the major synaptic vesicle protein p38) is a protein encoded by the SYP gene in humans. It is expressed in paneth cells, in goblet cells (in the mucus and cytoplasm) and in the neurons/synapses in the mucosa and submucosa. The gene plays a critical role in regulating neurotransmitter release and synaptic plasticity. It may be primarily associated with attention-deficit/hyperactivity disorder (ADHD). The gene is found to be involved in the susceptibility to schizophrenia. Abnormal expression of this gene in different brain regions may be linked to disorder in postmortem brain.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74355

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Liu et al.
Psychiatry research, 210(1), 308-314 (2013-06-04)
Dysfunction of neurotransmitters has been suggested to be involved in the etiology of attention-deficit/hyperactivity disorder (ADHD). Hence, genes encoding proteins involved in the vesicular release process of those neurotransmitters are attractive candidates in ADHD genetics. One of these genes is
Carlos A Rubio
In vivo (Athens, Greece), 26(1), 135-138 (2012-01-03)
Paneth cells are known to produce lysozyme, α-defensins, phospholipase, matrilysin and guanylin. This study reports, for the first time, that duodenal Paneth cells may also produce the neuroendocrine peptide synaptophysin. Normal duodenal biopsies from 37 patients were immunostained for synaptophysin
Yu-Chih Shen et al.
Schizophrenia research, 137(1-3), 14-19 (2012-02-22)
Synaptophysin (SYP) has been shown to be critical for regulating neurotransmitter release and synaptic plasticity, a process thought to be disrupted in schizophrenia. In addition, abnormal SYP expression in different brain regions has been linked to this disorder in postmortem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service