Skip to Content
Merck
All Photos(6)

Key Documents

HPA002328

Sigma-Aldrich

Anti-MAOB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Amine oxidase [flavin-containing] B, Anti-MAO-B, Anti-Monoamine oxidase type B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

WRTMDDMGREIPSDAPWKAPLAEEWDNMTMKELLDKLCWTESAKQLATLFVNLCVTAETHEVSALWFLWYVKQCGGTTRIISTTNGGQERKFVGGSGQVSERIMDLLGDRVKLERPVIYIDQTRENVLVETLNHEMYEAKYVISAIPPTLGAPHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAOB(4129)

General description

Monoamine oxidase (MAO) is localized in the outer mitochondrial membrane. It exists in two forms MAOA and MAOB. It is expressed in specific zone of brain i.e. hypothalamus, hippocampus, brain stem and olfactory bulb.

Immunogen

Amine oxidase [flavin-containing] B recombinant protein epitope signature tag (PrEST)

Application

Anti-MAOB antibody produced in rabbit has been used for immunostaining and immunohistochemistry.

Biochem/physiol Actions

MAOB (monoamine oxidase B) is involved in the oxidative metabolism of neurotransmitters and dietary amines, such as dopamine, neuroactive, vasoactive amines and the Parkinsonism-producing neurotoxin 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP). It forms H2O2 during dopamine metabolism. It is associated with the cause of Parkinson′s disease (PD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82420

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Nakatome et al.
Biochemical and biophysical research communications, 247(2), 452-456 (1998-06-27)
Monoamine oxidase (MAO), which exists in two forms (MAOA and MAOB), plays an important role in the oxidative metabolism of neurotransmitters such as dopamine, and has been implicated in the etiology of Parkinson's disease (PD). Individual variations in the activity
Astroglial Responses to Amyloid-Beta Progression in a Mouse Model of Alzheimer?s Disease
Olsen M, et al.
Molecular Imaging and Biology : MIB : The Official Publication of the Academy of Molecular Imaging, 20(8) (2017)
Tianjie Pu et al.
Science advances, 10(6), eadi4935-eadi4935 (2024-02-09)
Prostate cancer (PC) develops in a microenvironment where the stromal cells modulate adjacent tumor growth and progression. Here, we demonstrated elevated levels of monoamine oxidase B (MAOB), a mitochondrial enzyme that degrades biogenic and dietary monoamines, in human PC stroma
Aberrant Tonic Inhibition of Dopaminergic Neuronal Activity Causes Motor Symptoms in Animal Models of Parkinson?s Disease
Heo JY, et al.
Current Biology, 30(2) (2020)
Seonmi Jo et al.
Nature medicine, 20(8), 886-896 (2014-06-30)
In Alzheimer's disease (AD), memory impairment is the most prominent feature that afflicts patients and their families. Although reactive astrocytes have been observed around amyloid plaques since the disease was first described, their role in memory impairment has been poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service