Skip to Content
Merck
All Photos(8)

Key Documents

HPA001671

Sigma-Aldrich

Anti-STAT3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Acute-phase response factor antibody produced in rabbit, Anti-Signal transducer and activator of transcription 3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 1:100- 1:250
immunofluorescence: 1-4 μg/mL
immunohistochemistry: 1:50- 1:200

immunogen sequence

GVTFTWVEKDISGKTQIQSVEPYTKQQLNNMSFAEIIMGYKIMDATNILVSPLVYLYPDIPKEEAFGKYCRPESQEHPEADPGSAAPYLKTKFICVTPTTCSNTIDLPMSPRTLDSLMQFGNNGEGAEPSAGGQFESLTFDMELTSECA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT3(6774)

Immunogen

Signal transducer and activator of transcription 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Signal transducers and activators of transcription (STAT3) acts as a transcription factor in signaling pathways. STAT3 binds to the interleukin-6 (IL-6). It is mainly associated with various critical functions in the immune system, including control of dendritic cell production, suppression of macrophage inflammatory signaling, and regulation of steady state and emergency granulopoiesis. In addition, STAT3 also participates as a cytokine-activated essential regulator in inflammatory T helper cell (THi) development. STAT3 is activated by the non-tyrosine kinase containing cytokine receptors as well as by growth factor receptors such as EGFR (epidermal growth factor receptor) and the Src family of kinases (SFKs). Mutation in STAT3 gene causes hyperimmunoglobulin E syndrome. STAT3 also have been reported for its tumor suppression characteristics.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77483

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

STAT3 signaling and the hyper-IgE syndrome.
David E Levy et al.
The New England journal of medicine, 357(16), 1655-1658 (2007-09-21)
S Akira et al.
Cell, 77(1), 63-71 (1994-04-08)
Acute-phase response factor (APRF) is a transcription factor that binds to the interleukin-6 (IL-6)-responsive elements identified in the promoters of various acute-phase protein genes. We report here the purification and cloning of APRF. APRF exhibits a 52.5% overall homology at
Xuexian O Yang et al.
The Journal of biological chemistry, 282(13), 9358-9363 (2007-02-06)
Interleukin-17 (IL-17)-producing helper T (TH) cells, named as TH(IL-17), TH17, or inflammatory TH (THi), have been recently identified as a novel effector lineage. However, how cytokine signals mediate THi differentiation is unclear. We found that IL-6 functioned to up-regulate IL-23R
Corinne M Silva
Oncogene, 23(48), 8017-8023 (2004-10-19)
The signal transducers and activators of transcription (STATs) were originally identified in the signaling pathway activated by the nontyrosine kinase containing cytokine receptors. The role of these STATs in hematopoietic cell signaling has been well described. In the case of
Monica Musteanu et al.
Gastroenterology, 138(3), 1003-1011 (2009-12-08)
The transcription factor signal transducer and activator of transcription 3 (Stat3) has been considered to promote progression and metastasis of intestinal cancers. We investigated the role of Stat3 in intestinal tumors using mice with conditional ablation of Stat3 in intestinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service