Skip to Content
Merck
All Photos(5)

Key Documents

HPA001637

Sigma-Aldrich

Anti-TJP1 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Tight junction protein 1 antibody produced in rabbit, Anti-Tight junction protein ZO-1 antibody produced in rabbit, Anti-Zona occludens 1 protein antibody produced in rabbit, Anti-Zonula occludens 1 protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ASSQPAKPTKVTLVKSRKNEEYGLRLASHIFVKEISQDSLAARDGNIQEGDVVLKINGTVTENMSLTDAKTLIERSKGKLKMVVQRDERATLLNVPDLSDSIHSANASERDDISEIQSLASDHSGR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TJP1(7082)

Looking for similar products? Visit Product Comparison Guide

General description

Tight junction protein 1/Zona occludens 1 protein (TJP1, ZO-1) is a transmembrane plaque protein belonging to the membrane-associated guanylate kinase (MAGUK) family of scaffold proteins. It consists of three PDZ (PSD-95/Discs large/ZO-1) domains, one SH3 (Src-homology 3) domain, and one GK (guanylate kinase) domain. Additionaly, it also contains a long proline-rich carboxyl terminal region and a ZU5 domain.

Immunogen

Tight junction protein ZO-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tight junction protein ZO-1 is a protein encoded by the TJP1 gene in humans and is mapped to chromosome 15q13. It is a membrane-scaffolding protein that plays an important role in maintaining tight-junction integrity, which is disrupted in many invasive cancers and intestinal diseases. It may act as a tumor-suppressor gene in GIST. It is an integral component of functional TJs and is regulated by p38δ. It shows a hypermethylation status in children with non-Hodgkin lymphoma (NHL) and the methylation level is positively correlated with the number of malignant cells in bone marrow. It may be useful as a novel molecular marker in early diagnosis, outcome assessment, prognostic evaluation and detection of minimal residual disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83051

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huijun Zhu et al.
Medical oncology (Northwood, London, England), 30(3), 631-631 (2013-07-04)
Zonula occludens-1 (ZO-1) is a membrane-scaffolding protein that plays an important role in maintaining tight-junction integrity, which is disrupted in many invasive cancers and intestinal diseases. However, the expression of ZO-1 in gastrointestinal stromal tumor (GIST) and its relationship with
Yu-Qiao Diao et al.
Zhongguo dang dai er ke za zhi = Chinese journal of contemporary pediatrics, 16(6), 619-623 (2014-06-14)
To investigate the methylation status of zonula occludens-1 (ZO-1) gene promoter and its clinical significance in children with stage IV non-Hodgkin lymphoma (NHL) and to provide a basis for further etiological study and early diagnosis of this disease. Fifty-five children
Elina Siljamäki et al.
Archives of dermatological research, 306(2), 131-141 (2013-07-17)
Increasing evidence has recognized tight junctions (TJs) as the lower epidermal inside-out diffusion barrier located in granular cell layers of the epidermis. However, little is known about the regulation of TJ components in epidermis. p38 pathway is one of the
Fan Yang et al.
Frontiers in oncology, 11, 683367-683367 (2021-07-06)
Glioblastoma (GBM) is the most aggressive and lethal type of brain tumors. Magnetic resonance imaging (MRI) has been commonly used for GBM diagnosis. Contrast enhancement (CE) on T1-weighted sequences are presented in nearly all GBM as a result of high
Lin Huo et al.
The EMBO journal, 30(4), 665-678 (2011-01-18)
Zonula occludens (ZO)-1 is a multi-domain scaffold protein known to have critical roles in the establishment of cell-cell adhesions and the maintenance of stable tissue structures through the targeting, anchoring, and clustering of transmembrane adhesion molecules and cytoskeletal proteins. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service