Skip to Content
Merck
All Photos(2)

Key Documents

HPA001196

Sigma-Aldrich

Anti-AMOTL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Angiomotin-like protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SPSACYSPSSPVQVLEDSTYFSPDFQLYSGRHETSALTVEATSSIREKVVEDPLCNFHSPNFLRISEVEMRGSEDAAAGTVLQRLIQEQLRYGTPTENMNLLAIQHQATGSAGPAHPTNNFSSTENLTQEDPQMVYQSARQEPQGQEHQV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AMOTL1(154810)

Immunogen

Angiomotin-like protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-AMOTL1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

AMOTL1 (angiomotin like 1) encodes a peripheral membrane protein that forms a component of protein complex containing p80-angiomotin that promotes angiogenesis. It associates with p80-angiomotin-containing complex via its coiled-coil domain and plays a role in remodeling of the actin cytoskeleton. This protein interacts with M protein of parainfluenza virus 5 (PIV5) and plays a role in the viral infection. It negatively regulates the transcription factors YAP (Yes-associated protein) and TAZ (transcriptional coactivator with PDZ-binding motif) by preventing their nuclear translocation. It regulates endothelial polarity and tight junction stability during sprouting angiogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77462

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanjin Hong
Science signaling, 6(291), pe27-pe27 (2013-09-05)
The Hippo pathway regulates cell proliferation and apoptosis during development, tissue regeneration, and carcinogenesis. Nuclear translocation of the transcription factors Yes-associated protein (YAP) and transcriptional coactivator with PDZ-binding motif (TAZ) and their subsequent interaction with TEA domain (TEAD) transcriptional factors
Valérie Gagné et al.
Cell motility and the cytoskeleton, 66(9), 754-768 (2009-07-01)
Angiostatin is a potent inhibitor of angiogenesis. One mechanism through which angiostatin inhibits angiogenesis is by binding to the cell surface protein p80-angiomotin. The p80-angiomotin protein promotes angiogenesis, in part, by conferring a hypermigratory phenotype to endothelial cells. Although p80-angiomotin
Yujuan Zheng et al.
Circulation research, 105(3), 260-270 (2009-07-11)
We have previously shown that angiomotin (Amot) is essential for endothelial cell migration during mouse embryogenesis. However, approximately 5% of Amot knockout mice survived without any detectable vascular defects. Angiomotin-like protein 1 (AmotL1) potentially compensates for the absence of Amot
Zifei Pei et al.
Virology, 397(1), 155-166 (2009-11-26)
Paramyxovirus matrix (M) proteins organize virus assembly, functioning as adapters that link together viral ribonucleoprotein complexes and viral glycoproteins at infected cell plasma membranes. M proteins may also function to recruit and manipulate host factors to assist virus budding, similar
Anne E Roehrig et al.
PloS one, 16(8), e0254697-e0254697 (2021-08-24)
The PAF complex (PAFC) coordinates transcription elongation and mRNA processing and its CDC73/parafibromin subunit functions as a tumour suppressor. The NF2/Merlin tumour suppressor functions both at the cell cortex and nucleus and is a key mediator of contact inhibition but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service