Skip to Content
Merck
All Photos(4)

Documents

HPA000923

Sigma-Aldrich

Anti-THSD7A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Thrombospondin type-1 domain-containing protein 7A precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000-1:2500

immunogen sequence

VCQSSPCEAEELRYSLHVGPWSTCSMPHSRQVRQARRRGKNKEREKDRSKGVKDPEARELIKKKRNRNRQNRQENKYWDIQIGYQTREVMCINKTGKAADLSFCQQEKLPMTFQSC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... THSD7A(221981)

Immunogen

Thrombospondin type-1 domain-containing protein 7A precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76155

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fidelis Andrea Flockerzi et al.
International journal of molecular sciences, 24(13) (2023-07-14)
Lung cancer is the leading cause of cancer-related deaths in the western world, with squamous cell carcinoma being one of the most common histological subtypes. Prognostic and predictive markers are still largely missing for squamous cell carcinoma of the lung
Phillip R Stahl et al.
Genes, chromosomes & cancer, 56(4), 314-327 (2016-12-31)
We recently described a case of a Thrombospondin Type-1 Domain containing 7A (THSD7A) associated membranous nephropathy in a female patient who was synchronously suffering from a THSD7A-positive malignancy. We here investigated the role of THSD7A as a new potential tumor
Li Xian et al.
BMC nephrology, 20(1), 332-332 (2019-08-25)
Thrombospondin type 1 domain containing 7A (THSD7A) was recently identified target autoantigen in membranous nephropathy (MN). However, patients with positive THSD7A expression were prone to have malignancies. THSD7A was found to be expressed in a variety of malignant tumors. In
Fidelis Andrea Flockerzi et al.
Diagnostics (Basel, Switzerland), 13(2) (2023-01-22)
Prostate cancer is one of the most common malignancies, and there are a wide range of treatment options after diagnosis. Most prostate cancers behave in an indolent manner. However, a given sub-group has been shown to exhibit aggressive behavior; therefore
M Fresquet et al.
Journal of autoimmunity, 106, 102308-102308 (2019-08-10)
Patients with membranous nephropathy have autoantibodies against PLA2R (up to 80%), or THSD7A (up to 2%). We previously described the immunodominant epitope within PLA2R but epitopes in THSD7A are still unknown. To find anti-THSD7A sera for this study, we screened

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service