Skip to Content
Merck
All Photos(1)

Key Documents

H8909

Sigma-Aldrich

HBcAg core human

~1 mg/mL, ≥95% (SDS-PAGE), recombinant, expressed in E. coli

Synonym(s):

Hepatitis B virus core antigen

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

mol wt

18 kDa

concentration

~1 mg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

hepatitis B virus ... HBVgp4(944568)
human ... HBVgp4(944568)

Related Categories

General description

amino acids 1-183 of HBV core antigen.

Biochem/physiol Actions

HBcAg core human is a capsid protein of hepatitis B virus (HBV). It acts as a viral factor for HBV clearance. HBcAg is implicated in viral replication.
Hepatitis B virus core antigen (HBcAg) is a positive control for human HBV antibodies.In humans with chronic HBV (hepatitis B virus) infection, cytotoxic effects of blood T lymphocytes can be suppressed by incubating cells with antibodies against hepatitis B core antigen (HBcAg). The antigen can also work as a T-cell independent antigen in athymic mice. However, it is an efficient immunogen, resulting in T-cell activation and increased anti-HBc levels. High cytoplasmic/membranous expression of HBcAg is associated with increased liver inflammation.

Physical form

Solution in 50% glycerol, 7.5 mM phosphate buffer, pH 7.2, 75 mM NaCl.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hepatitis B virus nucleocapsid but not free core antigen controls viral clearance in mice
Lin YJ, et al.
Journal of Virology, 86(17), 9266-9273 (2012)
M Mondelli et al.
Journal of immunology (Baltimore, Md. : 1950), 129(6), 2773-2778 (1982-12-01)
Peripheral blood T lymphocytes from 21 patients with chronic HBV infection were incubated with autologous hepatocytes in a microcytotoxicity assay. Cytotoxicity was significantly increased in 13 cases, and in 12 of these the cytotoxic effect of the T lymphocytes was
D R Milich et al.
Journal of immunology (Baltimore, Md. : 1950), 139(4), 1223-1231 (1987-08-15)
Hepatitis B virus nucleocapsid particles (HBcAg) can function as a T cell-independent antigen when injected into athymic mice. However, immunization of euthymic mice with HBcAg results in dramatically increased anti-HBc titers. Therefore we have examined the murine T cell response
Jun Hong et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 38(4), 362-373 (2007-11-21)
This study was undertaken to investigate whether plasmacytoid dendritic cells (PDC) are involved in the generation of a higher proportion of CD4(+) and CD25(+) regulatory T (Treg) cells in chronic hepatitis B virus (HBV) infection compared with healthy patients. The
J B Kurtz et al.
Journal of virological methods, 96(2), 127-132 (2001-07-11)
A measurable serological response to hepatitis C infection is delayed on average until 70 days after infection. In addition, it may not occur in some immunocompromised people. Detection of free hepatitis C (HCV) core antigen in blood has enabled diagnosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service