Skip to Content
Merck
All Photos(1)

Key Documents

H8891

Sigma-Aldrich

Heparinase III from Flavobacterium heparinum

Lyophilized powder stabilized with approx. 25% (w/w) bovine serum albumin, ≥30 units/mg protein (enzyme + BSA)

Synonym(s):

Heparin Lyase III, Heparitinase I, Heparitinase from Flavobacterium heparinum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Flavobacterium heparinum)

Quality Level

conjugate

conjugate (Glucosaminoglycan)

form

lyophilized powder

specific activity

≥30 units/mg protein (enzyme + BSA)

mol wt

70.8 kDa

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Heparinase III from Flavobacterium heparinum has been used to cleave specific GAGs. It has been used to study its effect on the electrophoretic mobility of the protein reacting with the anti-aFGF (acidic fibroblast growth factor) antibody.

Biochem/physiol Actions

Heparin-degrading lyase that recognizes heparin sulfate proteoglycan as its primary substrate.
Heparinase III prevents the formation of new blood vessels in vivo. It specifically degrades heparin sulphate at the HNY-I and HNY,6X-G2 links. Unlike other heparinise, heparinase III does not contain the amino acid cysteine. It contains 13 histidine residues, which is responsible for its catalytic action. This enzyme is not active towards heparin or low molecular weight heparins.

Unit Definition

One unit will form 0.1 μmole of unsaturated uronic acid per hr at pH 7.5 at 25 °C. One International Unit (I.U.) is equivalent to approx. 600 Sigma units. Package sizes are sold in Sigma units.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composition of the endothelial glycocalyx and its relation to its thickness and diffusion of small solutes
Gao L and Lipowsky HH
Microvascular Research, 80(3), 394-401 (2010)
Andrea Garolla et al.
Human reproduction (Oxford, England), 27(4), 967-973 (2012-02-09)
Human papillomavirus (HPV) infection has been demonstrated in the sperm of a large percentage of sexually active males and is associated with an impairment of sperm parameters, with a particular negative impact on sperm motility, suggesting a possible role in
A Constantinescu et al.
Thrombosis and haemostasis, 105(5), 790-801 (2010-12-22)
A thick endothelial glycocalyx contributes to the barrier function of vascular endothelium in macro- and microcirculation. We hypothesised in the current study that diet-induced hyperlipidaemia perturbs the glycocalyx, resulting in decreased dimensions of this layer and increased transendothelial lipoprotein leakage
Md Nasimuzzaman et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 20(6), 1158-1166 (2012-03-22)
Foamy viruses (FVs) (spumaretroviruses) are good alternative to retroviruses as gene therapy vector. Despite four decades since the discovery of FV, its receptor molecule is still unknown. FV vector transduction of human CD34(+) cells was inhibited by culture with fibronectin.
FGF binding by extracellular matrix components of Wharton's jelly
Malkowski A, et al.
Acta Biochimica Polonica, 54(2), 357-357 (2007)

Articles

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

Uncover more about glycosaminoglycans and proteoglycans including the structure of glycosaminoglycans (GAGs), the different types of GAGs, and their functions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service