Skip to Content
Merck
All Photos(6)

Documents

H0131

Sigma-Aldrich

DL-4-Hydroxy-3-methoxymandelic acid

≥98% (HPLC), powder

Synonym(s):

α,4-Dihydroxy-3-methoxybenzeneacetic acid, VMA, Vanillomandelic acid, Vanillylmandelic acid, Vanilmandelic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH(OH)CO2H
CAS Number:
Molecular Weight:
198.17
Beilstein:
2213227
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

132-134 °C (dec.) (lit.)

solubility

H2O: ~50 mg/mL

storage temp.

2-8°C

SMILES string

OC1=C(OC)C=C(C(O)C(O)=O)C=C1

InChI

1S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11H,1H3,(H,12,13)

InChI key

CGQCWMIAEPEHNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Epinephrine and norepineprine metabolite.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Garnier et al.
Journal of pediatric surgery, 47(8), e21-e23 (2012-08-21)
Mesoblastic nephroma is by far the most frequent intrarenal fetal tumor. To the best of our knowledge, we report the first case of a newborn with an intrarenal neuroblastoma that was discovered prenatally. An intrarenal echogenic and homogenous mass was
W P Tormey et al.
Irish journal of medical science, 164(2), 146-150 (1995-04-01)
A study of the case notes from 101 of the 113 instances where at least one of the results was abnormal from the analysis of 24 hr urinary collections for noradrenaline, adrenaline, and dopamine and their metabolites 4-hydroxy-3-methoxy-mandelic acid (HMMA)
R Krishnappa et al.
Indian journal of cancer, 49(1), 188-193 (2012-07-31)
To review the clinical experience on pheochromocytoma in Indian subset of patients. Ten patients diagnosed with pheochromocytoma between 2001 and 2010 at our institute were retrospectively studied for clinical, laboratory, radiological and surgical data. A total of 10 patients (5
Wayne L Furman et al.
Investigational new drugs, 30(4), 1660-1670 (2011-07-29)
Gefitinib potently inhibits neuroblastoma proliferation in vitro, and the gefitinib/irinotecan combination shows greater than additive activity against neuroblastoma xenografts. This Phase II pilot study estimated the rate of response to two courses of intravenous irinotecan plus oral gefitinib in children
Kai Li et al.
Journal of investigative surgery : the official journal of the Academy of Surgical Research, 25(2), 86-92 (2012-03-24)
This study assesses the clinical features of neuroblastoma and survival. Data for 98 patients between January 2000 and December 2006 at Children's Hospital of Fudan University, Shanghai, China, were retrospectively analyzed. Diagnostic methods included imaging, 24-hr urine catecholamines, bone marrow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service