Skip to Content
Merck
All Photos(2)

Documents

GHS132

Sigma-Aldrich

Hematoxylin Solution, Gill No. 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

2 g/L

pH

2.5-3.5

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gill No. 1 formulation is used as a progressive cytology stain. Used with hematoxylin and eosin staining.

Other Notes

2 g/L certified hematoxylin

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J De Andrés et al.
Reproduction in domestic animals = Zuchthygiene, 51(2), 269-275 (2016-02-24)
Canine inflammatory mammary cancer (IMC) has been proposed as a model for the study of human inflammatory breast cancer (IBC). The aims of this study were to compare the immunohistochemical expression of aromatase (Arom) and several hormone receptors [estrogen receptor
Tiziana Annese et al.
Histochemistry and cell biology, 153(3), 185-192 (2019-12-18)
Diffuse large B-cell lymphomas (DLBCLs) are the most common types of Non-Hodgkin's lymphomas and are highly heterogeneous in terms of phenotype and treatment response. The natural course of DLBCLs tumor progression is featured by a flow of events leading to
Nathan Ungerleider et al.
PLoS pathogens, 14(8), e1007206-e1007206 (2018-08-07)
Our appreciation for the extent of Epstein Barr virus (EBV) transcriptome complexity continues to grow through findings of EBV encoded microRNAs, new long non-coding RNAs as well as the more recent discovery of over a hundred new polyadenylated lytic transcripts.
Arinjay Banerjee et al.
Scientific reports, 10(1), 7257-7257 (2020-05-01)
Coronaviruses that cause severe acute respiratory syndrome (SARS) and Middle East respiratory syndrome (MERS) are speculated to have originated in bats. The mechanisms by which these viruses are maintained in individuals or populations of reservoir bats remain an enigma. Mathematical
Patrick P van Vliet et al.
Developmental biology, 429(1), 249-259 (2017-07-04)
Correct cardiac development is essential for fetal and adult life. Disruptions in a variety of signaling pathways result in congenital heart defects, including outflow and inflow tract defects. We previously found that WNT11 regulates outflow tract development. However, tissue specific

Related Content

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Brief information on the field of cytology, its role in cytodiagnosis and the use of standard and special stains for microscopic analysis to detect cellular abnormalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service