GENA934-100UL
Rabbit IgG HRP Linked Whole Ab (from Donkey)
Cytiva NA934-100UL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.76
feature
wetted part: no
packaging
pkg of 100 μL
manufacturer/tradename
Cytiva NA934-100UL
color
Brown (light)
shipped in
wet ice
storage temp.
2-8°C
Related Categories
General description
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2° C to 8° C.
Features and Benefits
- Highly species-specific.
- Optimized for use with our range of ECL™ Detection Reagents.
- Recommended dilutions to minimize background.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
ECL is a trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service