GE10600053
Amersham™ Protran® Supported Western blotting membranes, nitrocellulose
pore size 0.2 μm, sheet W × L 200 mm × 200 mm, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
packaging
pkg of 25 ea
manufacturer/tradename
Cytiva 10600053
sheet W × L
200 mm × 200 mm
pore size
0.2 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Amersham™ Protran® Supported Western blotting membranes, nitrocellulose, a robust protein blotting membrane, is made of reinforced nitrocellulose for multiple reprobings.
Application
Amersham™ Protran® Supported Western blotting membranes, nitrocellulose is suitable for use in:
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- High binding capacity.
- Versatile, robust and excellent signal-to-noise ratios.
- Optimal for applications when stripping and reprobing is required.
- Suitable for proteins of a wide range of molecular weights.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service