Skip to Content
Merck
All Photos(5)

Key Documents

G8634

Sigma-Aldrich

Guanosine 5′-[γ-thio]triphosphate tetralithium salt

≥90% (contains < 10% GDP, HPLC), powder

Synonym(s):

GTPγS tetralithium salt, GTP-γ-S-Li4, Guanosine 5′-O-(3-thiotriphosphate) tetralithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12Li4N5O13P3S
CAS Number:
Molecular Weight:
562.98
Beilstein:
8182249
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥90% (contains < 10% GDP, HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥75 mg/mL (Solutions are very unstable; prepare immediately prior to use.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].[Li+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=S)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H16N5O13P3S.4Li/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(26-9)1-25-29(19,20)27-30(21,22)28-31(23,24)32;;;;/h2-3,5-6,9,16-17H,1H2,(H,19,20)(H,21,22)(H2,23,24,32)(H3,11,13,14,18);;;;/q;4*+1/p-4/t3-,5-,6-,9-;;;;/m1..../s1

InChI key

AMQXJFWJOAWCPV-ZVQJTLEUSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Guanosine 5′-[γ-thio]triphosphate tetralithium salt has been used:
  • as a component of lysis buffer
  • in oocyte and mammalian cell patch-clamp experiments to keep the G proteins in a constantly active state
  • to couple with G- protein α, to study its effect on kinase activity

Biochem/physiol Actions

GTP-γ-S is a non-hydrolyzable GTP analog, known for its role as a G-protein activator. It has recently been shown to protect proteins from proteolytic degradation, stimulate GLUT4 translocation in a tyrosine kinase-dependent manner, stimulate phospholipases and induce actin polymerization in vitro.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rab27a is an essential component of melanosome receptor for myosin Va
Wu X, et al.
Molecular Biology of the Cell, 13(5), 1735-1749 (2002)
Brain-specific Galphaz interacts with Src tyrosine kinase to regulate Mu-opioid receptor-NMDAR signaling pathway
Sanchez BP, et al.
Cellular Signalling, 21(9), 1444-1454 (2009)
Jing Geng et al.
Nature communications, 12(1), 3519-3519 (2021-06-12)
TLR4 signaling plays key roles in the innate immune response to microbial infection. Innate immune cells encounter different mechanical cues in both health and disease to adapt their behaviors. However, the impact of mechanical sensing signals on TLR4 signal-mediated innate
Chantal Binda et al.
The Journal of biological chemistry, 294(45), 16865-16883 (2019-10-03)
Accumulating evidence indicates that G protein-coupled receptors (GPCRs) interact with Rab GTPases during their intracellular trafficking. How GPCRs recruit and activate the Rabs is unclear. Here, we report that depletion of endogenous L-type prostaglandin D synthase (L-PGDS) in HeLa cells
Trine Ljungstrom et al.
Pflugers Archiv : European journal of physiology, 446(6), 684-694 (2003-06-27)
Activation of KCNQ potassium channels by stimulation of co-expressed dopamine D(2) receptors was studied electrophysiologically in Xenopus laevis oocytes and in mammalian cells. To address the specificity of the interaction between D(2)-like receptors and KCNQ channels, combinations of KCNQ1-5 channels

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service