G8546
p38γ, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
ERK3, ERK6, MAPK12, PRKM12, SAPK3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51111800
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
56-76 nmol/min·mg
mol wt
~71 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... MAPK12(6300)
General description
p38γ, commonly known as MAPK12 (mitogen-activated protein kinase 12), is one of the four isoforms of the p38 MAPK family. This isoform shows predominant expression in skeletal muscle and is controlled by exercise.
Biochem/physiol Actions
p38γ is a key protein of the MAPK (mitogen-activated protein kinase) pathway and is involved in cell differentiation, growth, cycle and apoptosis and inflammation. It is essential for normal localization of polo-like kinase 1 (Plk1) at the kinetochore. This protein facilitates mitotic cell viability and suppresses chromosomal instability. Studies suggest that this protein negatively controls the expression of GLUT4 (glucose transporter) and the uptake of glucose mediated by contraction in adult skeletal muscles.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anu Kukkonen-Macchi et al.
Journal of cell science, 124(Pt 2), 216-227 (2010-12-22)
The p38 mitogen-activated protein kinase (p38 MAPK) family, which is comprised of four protein isoforms, p38α, p38β, p38γ and p38δ, forms one of the key MAPK pathways. The p38 MAPKs are implicated in many cellular processes including inflammation, differentiation, cell
Richard C Ho et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 286(2), R342-R349 (2003-11-01)
Skeletal muscle expresses at least three p38 MAPKs (alpha, beta, gamma). However, no studies have examined the potential regulation of glucose uptake by p38gamma, the isoform predominantly expressed in skeletal muscle and highly regulated by exercise. L6 myotubes were transfected
Z Li et al.
Biochemical and biophysical research communications, 228(2), 334-340 (1996-11-12)
We have identified a third member of the p38 group of MAP kinase termed p38 gamma. The cDNA for this MAP kinase encodes an 367 amino acid polypeptide that is slightly greater than 60% identical to p38 and p38 beta.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service