Skip to Content
Merck
All Photos(3)

Key Documents

G4046

Sigma-Aldrich

Anti-Glucocerebrosidase antibody produced in rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-D-glucosyl-N-acylsphingosine glucohydrolase, Anti-GBA, Anti-GBA1, Anti-Glucosidase, beta (Gluc), Anti-Glucosylceramidase (GlcCerase), Anti-Lysosomal glucocerebrosidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

mouse, human, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cell lysate expressing human glucocerebrosidase (GBA) or SH-SY-5Y cell lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GBA(2629)
mouse ... Gba(14466)

General description

GBA1 (β-glucocerebrosidase) gene is mapped to human chromosome 1q21. It is a lysosomal enzyme and is widely expressed. The protein has domain I with three stranded anti?parallel βsheets, domain II with two β sheets making an immunoglobulin like domain and domain III with eight stranded β/α triosephosphate isomerase (TIM) barrel.

Application

Anti-Glucocerebrosidase antibody produced in rabbit has been used in western blotting and immunocytochemistry.
Anti-Glucocerebrosidase antibody produced in rabbit is suitable for immunoblotting at a working concentration of 1-2μg/mL using HEK-293T cell lysate expressing human glucocerebrosidase (GBA).

Biochem/physiol Actions

GBA (glucosidase, beta, acid) gene encodes a protein that cleaves the β-glucosidic linkage of glycosylceramide (GlcCer), an intermediate in glycolipid metabolism. Mutations in the human GBA gene cause a reduction in the GBA activity and accumulation of GlcCer in lysosomes of cells of the reticuloendothelial system. This causes Gaucher disease (GD), an inherited lysosomal storage disorder, characterised by severe loss of neurons in the central nervous system, fetal onset, hydrops fetalis, in utero fetal death and neonatal distress. It leads to changes in neuronal functionality including increased levels of tubular endoplasmic reticulum (ER) elements, a large increase in Ca2+ release from the ER in response to glutamate, and an increased sensitivity to glutamate-induced neurotoxicity. Mutations in the human GBA gene may contribute to the development of common age-related dementia known as dementia with Lewy bodies or DLB and may cause early-onset of Parkinson disease.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The relationship between glucocerebrosidase mutations and Parkinson disease
Migdalska RA, et al.
Journal of Neurochemistry, 1-1 (2016)
K P Zimmer et al.
The Journal of pathology, 188(4), 407-414 (1999-08-10)
Gaucher's disease (GD) is caused by an inherited deficiency of acid beta-glucosidase with storage of glucosylceramides in the lysosomes of macrophages. This study identifies a G202R mutation in the acid beta-glucosidase gene in an infant with severe neuronopathic (type 2)
E Korkotian et al.
The Journal of biological chemistry, 274(31), 21673-21678 (1999-07-27)
Gaucher disease is a glycosphingolipid storage disease caused by defects in the activity of the lysosomal hydrolase, glucocerebrosidase (GlcCerase), resulting in accumulation of glucocerebroside (glucosylceramide, GlcCer) in lysosomes. The acute neuronopathic type of the disease is characterized by severe loss
Dermal fibroblasts from patients with Parkinson?s disease have normal GCase activity and autophagy compared to patients with PD and GBA mutations
Collins LM, et al.
F1000Research (2017)
Individualized screening for chaperone activity in Gaucher disease using multiple patient derived primary cell lines
Ivanova MM, et al.
American Journal of Translational Research, 10(11), 3750-3750 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service