Skip to Content
Merck
All Photos(1)

Key Documents

G3665

Sigma-Aldrich

β-Galactosidase from Kluyveromyces lactis

≥2600 units/g

Synonym(s):

Lactozyme® 2600 L, Lactase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

liquid

Quality Level

specific activity

≥2600 units/g

concentration

≥2600 units/g

storage temp.

2-8°C

General description

A β-galactosidase preparation produced by submerged fermentation of a selected strain of the yeast Kluyveromyces lactis.

Application

β-galactosidases from different species, including Kluyveromyces lactis and Kluyveromyces fragilis, hydrolyze lactose.

Biochem/physiol Actions

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyzes the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.

Legal Information

A product of Novozyme Corp.
Lactozyme is a registered trademark of Novozymes Corp.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert E Ward et al.
Molecular nutrition & food research, 51(11), 1398-1405 (2007-10-30)
This study was conducted to investigate the catabolism and fermentation of human milk oligosaccharides (HMO) by individual strains of bifidobacteria. Oligosaccharides were isolated from a pooled sample of human milk using solid-phase extraction, and then added to a growth medium
Arun Beniwal et al.
3 Biotech, 7(5), 349-349 (2017-09-29)
The dairy yeast
Justyna Skoczek et al.
Food chemistry, 294, 231-237 (2019-05-28)
A fully mechanized Arduino-controlled multi-pumping flow analysis system and procedure for the determination of β-galactosidase activity are proposed. The applied bioanalytical method is based on the determination of p-nitrophenol formed in the course of enzyme-catalyzed hydrolysis of p-nitrophenyl-galactopyranosides. The photometric
D H Juers et al.
Protein science : a publication of the Protein Society, 8(1), 122-136 (1999-04-21)
Beta-galactosidase (lacZ) from Escherichia coli is a 464 kDa homotetramer. Each subunit consists of five domains, the third being an alpha/beta barrel that contains most of the active site residues. A comparison is made between each of the domains and
Anca D Petrescu et al.
Protein expression and purification, 58(2), 184-193 (2008-01-08)
Acyl coenzyme A binding protein (ACBP) has been proposed to transport fatty acyl CoAs intracellularly, facilitating their metabolism. In this study, a new mouse recombinant ACBP was produced by insertion of a histidine (his) tag at the C-terminus to allow

Articles

For use as a marker in SDS-PAGE; Albumin from chicken egg white, For use as a marker in SDS-PAGE; L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service