Skip to Content
Merck
All Photos(2)

Key Documents

G3651

Sigma-Aldrich

α-Glucosidase from Bacillus stearothermophilus

greener alternative

lyophilized powder, ≥50 units/mg protein

Synonym(s):

α-D-Glucoside glucohydrolase, Maltase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

specific activity

≥50 units/mg protein

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

foreign activity

β-Glucosidase, α−galactosidase and β−galactosidase ≤0.1%

greener alternative category

relevant disease(s)

diabetes (management of non-insulin-dependent diabetes mellitus)

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch hydrolysis research. For more information see the article in biofiles.

Application

α-glucosidase is potential enzyme for the biosynthesis of complex carbohydrates.[1]

Biochem/physiol Actions

α-glucosidase hydrolyzes carbohydrates by acting on 1,4-α linkages. Inhibition of α-glucosidase is a prominent target in the management of non-insulin-dependent diabetes mellitus[2].

Unit Definition

One unit will liberate 1.0 μmole of D-glucose from p-nitrophenyl α-D-glucoside per min at pH 6.8 at 37 °C.

Physical form

Lyophilized powder containing potassium phosphate buffer salt

Analysis Note

Activity using maltose as substrate at pH 6.0 at 25 deg C is ~2X greater than that obtained using p-nitrophenyl-α-D-glucoside as substrate at pH 6.8 at 37 °C.
Protein determined by biuret.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Setlow et al.
Journal of applied microbiology, 120(5), 1326-1335 (2016-01-26)
This study was to determine the sources, location and identity of α-glucosidases in dormant/germinating/outgrowing spores and growing cells of Geobacillus stearothermophilus ATCC 7953, an enzymatic activity in spores used in rapid tests of steam sterilization. α-Glucosidase activity in spores and
Arzu Coleri Cihan et al.
Cell biochemistry and function, 30(1), 69-81 (2011-10-27)
Two different α-glucosidase-producing thermophilic E134 strains were isolated from a hot spring in Kozakli, Turkey. Based on the phenotypic, phylogenetic and chemotaxonomic evidence, the strain was proposed to be a species of G. toebii. Its thermostable exo-α-1,4-glucosidases also were characterized
Sanaid Ghauri et al.
3 Biotech, 11(4), 167-167 (2021-04-06)
Diabetes mellitus is a metabolic disorder of carbohydrate metabolism. The management of Diabetes mellitus with phytochemicals is hallmark of this research. Citrus species are known for their health benefits and are used as traditional food in South East Asia. The
A D Baron
Diabetes research and clinical practice, 40 Suppl, S51-S55 (1998-09-18)
Fasting blood glucose level is usually used to diagnose diabetes, but is not a good predictor of postprandial hyperglycaemia, which is a more accurate measure of the metabolic defect underlying type 2 diabetes. Postprandial blood glucose levels may be elevated
Jenna Magat et al.
The Journal of biological chemistry, 298(12), 102625-102625 (2022-10-29)
Mucopolysaccharidosis type IIIA (MPS IIIA) is a lysosomal storage disorder caused by N-sulfoglucosamine sulfohydrolase (SGSH) deficiency. SGSH removes the sulfate from N-sulfoglucosamine residues on the nonreducing end of heparan sulfate (HS-NRE) within lysosomes. Enzyme deficiency results in accumulation of partially

Questions

1–3 of 3 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  3. What is the molecular weight of Product G3651, α-Glucosidase from Bacillus stearothermophilus?

    1 answer
    1. The molecular weight of Product G3651, α-Glucosidase from Bacillus stearothermophilus, enzyme is 50,000 daltons.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service