Skip to Content
Merck
All Photos(1)

Key Documents

G2904

Sigma-Aldrich

Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse

clone GT-34, ascites fluid

Synonym(s):

Monoclonal Anti-Goat/Sheep IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

GT-34, monoclonal

contains

15 mM sodium azide

species reactivity

sheep, goat, bovine

should not react with

rat, canine, mouse, rabbit, guinea pig, chicken, horse, pig, feline, human

technique(s)

indirect ELISA: 1:10,000-1:20,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG is the main type of antibody in human serum, secreted by the B cells. It is found in blood. IgG consists of four polypeptide chains, made of two identical 50 kDa γ heavy (H) chains and two identical 25 kDa κ or λ light (L) chains, which are linked by inter-chain disulfide bonds. IgG contains four isotypes IgG1, IgG2, IgG3 and IgG4.
The antibody reacts with an epitope located on the heavy chain of goat IgG1 and IgG2.

Specificity

Monoclonal Anti-Goat/Sheep IgG recognizes an epitope located on the heavy chain of goat IgG1 and IgG2. The antibody shows strong cross-reactivity with sheep IgG and weak cross-reactivity with bovine IgG. No cross-reactivity is observed with IgG derived from human, guinea pig, rat, horse, dog, chicken, pig, mouse, rabbit, or cat.

Application

Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunoprecipitation
  • immunoelectrophoresis

Biochem/physiol Actions

IgG stimulates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a crucial role in antibody-dependent cell-mediated cytotoxicity (ADCC). IgG might be associated with allergy. It has the longest serum half life.

Physical form

The product is provided as ascites fluid with 0.1% sodium azide as preservative.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seasonal variations of phagocytic response, immunoglobulin G (IgG) and plasma cortisol levels in Dhofari goats
Al-Busaidi R, et al.
Small Ruminant Research, 79(2-3), 118-123 (2008)
Tom N McNeilly et al.
Veterinary research, 44, 70-70 (2013-08-24)
Teladorsagia circumcincta is an important pathogenic nematode of sheep. It has been demonstrated previously that stimulation of murine T lymphocytes with excretory-secretory (ES) products derived from fourth stage larvae of T. circumcincta (Tci-L4-ES) results in de novo expression of Foxp3
Guillaume Sallé et al.
International journal for parasitology, 48(5), 395-402 (2018-03-15)
Some nematode species are economically important parasites of livestock, while others are important human pathogens causing some of the most important neglected tropical diseases. In both humans and animals, anthelmintic drug administration is the main control strategy, but the emergence
Su Ling Leong et al.
Current biology : CB, 29(13), 2199-2207 (2019-07-10)
Microtubule (MT) nucleation depends on the γ-tubulin complex (γ-TuC), in which multiple copies of the heterotetrameric γ-tubulin small complex (γ-TuSC) associate to form a ring-like structure (in metazoans, γ-tubulin ring complex; γ-TuRC) [1-7]. Additional conserved regulators of the γ-TuC include
Reconstitution of Microtubule Nucleation InA Vitro Reveals Novel Roles for Mzt1.
Leong, et al.
Current Biology, 33, 1864-1864 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service