Skip to Content
Merck
All Photos(2)

Key Documents

G2536

Sigma-Aldrich

Ganciclovir

≥99% (HPLC), powder, viral DNA elongation inhibitor

Synonym(s):

2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, 9-(1,3-Dihydroxy-2-propoxymethyl)guanine, DHPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N5O4
CAS Number:
Molecular Weight:
255.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Ganciclovir, ≥99% (HPLC), powder

Quality Level

Assay

≥99% (HPLC)

form

powder

color

white

solubility

0.1 M HCl: 10 mg/mL

ε (extinction coefficient)

12.0 at 256 nm at 1 mM

antibiotic activity spectrum

viruses

Mode of action

DNA synthesis | interferes

originator

Roche

storage temp.

2-8°C

SMILES string

NC1=Nc2c(ncn2COC(CO)CO)C(=O)N1

InChI

1S/C9H13N5O4/c10-9-12-7-6(8(17)13-9)11-3-14(7)4-18-5(1-15)2-16/h3,5,15-16H,1-2,4H2,(H3,10,12,13,17)

InChI key

IRSCQMHQWWYFCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: nucleoside

Application

Ganciclovir is used in molecular biology for selection against random recombination events when homologous recombination of a gene of interest is required.

Biochem/physiol Actions

Ganciclovir is a pro-drug nucleoside analog that is activated by phosphorylation. It is useful in the study of gene therapy in cancer research.
Upon expression of a viral suicide gene encoding thymidine kinase, the non-toxic pro-drug is converted to a phosphorylated active analog and is incorporated into the DNA of replicating eukaryotic cells, causing death of the malignant dividing cell. The cell cycle is irreversibly arrested at the G2-M checkpoint. Gap junction involvement in the ganciclovir bystander effect has been studied. Ganciclovir has been used to study loss of telomeres and to evaluate sensitivity of viruses to antiviral treatments.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Ganciclovir is soluble in 0.1 M HCl (10 mg/ml), DMSO (5 mg/ml), water (2 mg/ml), hot methanol, and ethanol (<1 mg/ml).
This product should be stored desiccated at 2-8 °C. Under these conditions the product is stable for 3 years.
Ganciclovir is tested for solubility in 0.1M HCl at 10 mg/mL. The compound is also soluble in DMSO at 5 mg/mL.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Cates Scott et al.
Therapeutic drug monitoring, 26(1), 68-77 (2004-01-30)
The authors use a previously published decision-making algorithm to address the role of clinical pharmacokinetic monitoring of ganciclovir, the drug of choice for prophylaxis and treatment of cytomegalovirus (CMV) in solid organ transplant recipients. Ganciclovir pharmacokinetics have been studied in
Koenraad Smets et al.
European journal of pediatrics, 165(12), 885-890 (2006-06-21)
The objective of this study is to look for evidence based or scientific guidelines for selection of newborns with congenital cytomegalovirus (CMV) infection that might benefit from treatment with ganciclovir. A literature search was conducted involving the MEDLINE database and
Zhanguo Gao et al.
Biomolecules, 11(10) (2021-10-24)
Duchenne muscular dystrophy (DMD), caused by the loss of dystrophin, remains incurable. Reduction in muscle regeneration with DMD is associated with the accumulation of fibroadipogenic progenitors (FAPs) differentiating into myofibroblasts and leading to a buildup of the collagenous tissue aggravating
Sunwen Chou
Reviews in medical virology, 18(4), 233-246 (2008-04-03)
Mutations in the human CMV UL97 kinase gene are a major mechanism of viral resistance to two anti-CMV drugs, ganciclovir (GCV) and maribavir (MBV). GCV, the most widely used and established therapy for CMV, is a substrate for the UL97
Marisa Bowers et al.
Blood, 125(17), 2678-2688 (2015-03-07)
Hematopoietic stem cells (HSCs) reside in regulatory niches in the bone marrow (BM). Although HSC niches have been extensively characterized, the role of endosteal osteoblasts (OBs) in HSC regulation requires further clarification, and the role of OBs in regulating leukemic

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service