Skip to Content
Merck
All Photos(4)

Key Documents

G110

Sigma-Aldrich

Guanabenz acetate salt

powder

Synonym(s):

1-(2,6-Dichlorobenzylideneamino)guanidine acetate salt, WY-8678

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8Cl2N4 · C2H4O2
CAS Number:
Molecular Weight:
291.13
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

color

white

solubility

H2O: 10 mg/mL
alcohol: 50 mg/mL

originator

Wyeth

SMILES string

CC(O)=O.NC(=N)N\N=C\c1c(Cl)cccc1Cl

InChI

1S/C8H8Cl2N4.C2H4O2/c9-6-2-1-3-7(10)5(6)4-13-14-8(11)12;1-2(3)4/h1-4H,(H4,11,12,14);1H3,(H,3,4)/b13-4+;

InChI key

MCSPBPXATWBACD-GAYQJXMFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Centrally acting α2 adrenoceptor agonist; I2 imidazoline binding site ligand; antihypertensive.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the α2-Adrenoceptors and Imidazoline Binding Sites pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Déborah Tribouillard-Tanvier et al.
PloS one, 3(4), e1981-e1981 (2008-04-24)
Prion-based diseases are incurable transmissible neurodegenerative disorders affecting animals and humans. Here we report the discovery of the in vivo antiprion activity of Guanabenz (GA), an agonist of alpha2-adrenergic receptors routinely used in human medicine as an antihypertensive drug. We
Melissa J Fullwood et al.
Progress in molecular biology and translational science, 106, 75-106 (2012-02-22)
The unfolded protein response, also known as endoplasmic reticulum (ER) stress, has been implicated in numerous human diseases, including atherosclerosis, cancer, diabetes, and neurodegenerative disorders. Protein misfolding activates one or more of the three ER transmembrane sensors to initiate a
Pavel Tsaytler et al.
The FEBS journal, 280(2), 766-770 (2012-02-22)
Selective and reversible phosphorylation is one of the most common post-translational modifications of proteins. Although kinase inhibitors are popular in drug development programmes, selective pharmacological manipulation of phosphatase activity has been challenging to achieve. We review recent advances in the
B Holmes et al.
Drugs, 26(3), 212-229 (1983-09-01)
Guanabenz is an orally active central alpha 2-adrenoceptor agonist. Its antihypertensive action is thought to result from a decrease in sympathetic outflow from the brain to the peripheral circulatory system as a result of stimulation of central alpha 2-adrenoceptors. In
Suzana Dos Reis et al.
Biochimie, 93(6), 1047-1054 (2011-03-15)
The ribosome, the protein synthesis machinery of the cell, has also been implicated in protein folding. This activity resides within the domain V of the main RNA component of the large subunit of the ribosome. It has been shown that

Articles

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service