Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF, IHC (p)
Citations:
8
biological source
rabbit
conjugate
FITC conjugate
antibody form
IgG fraction of antiserum
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:160, indirect immunofluorescence: 1:200
storage temp.
−20°C
target post-translational modification
unmodified
General description
The immunoglobulin G (IgG) is a major class of immunoglobulin and is highly conserved. IgG constitutes to 10−20% of the plasma proteins. This class of immunoglobulins are glycoproteins containing 82−96% protein and the remaining, carbohydrate. It exists in four subclasses namely, IgG1, IgG2, IgG3, and IgG4. IgG is composed of four polypeptide chains containing two heavy and two light chains, linked by disulfide bonds.
Application
Anti-Goat IgG (whole molecule)-FITC antibody produced in rabbit has been used for immunocytochemistry of human oocytes and preimplantation embryos.
It has also been used in enzyme linked immunosorbent assay (ELISA) and confocal microscopy.
Biochem/physiol Actions
Deficiency of a particular IgG subclass, may increase the susceptibility towards certain pathogens.
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of FITC to Anti- Goat IgG (whole molecule) antibody allows for the visualization of protein by fluorescent microscopy.
The antiserum is determined to be immunospecific for goat IgG.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Preparation Note
For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Goat IgG (whole molecule)–FITC antibody produced in rabbit
— or —
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Peter N Pushparaj et al.
Journal of immunology (Baltimore, Md. : 1950), 183(2), 1413-1418 (2009-07-01)
VAMP8, a member of the soluble N-ethylmaleimide-sensitive fusion protein attachment protein receptor (SNARE) family of fusion proteins, initially characterized in endosomal and endosomal-lysosomal fusion, may also function in regulated exocytosis. VAMP8 physiological function in inflammation has not been elucidated. In
29-kDa: A Potential Candidate for Anti-Tick Vaccine Antigen Source as Immunogenic and Stage Reactive Targeting Hard-Bodied Hyalomma Ticks (Ixodidae)
Kamran K, et al.
Indian Journal of Animal Sciences (India) , 7 null
G Cauffman et al.
Molecular human reproduction, 11(3), 173-181 (2005-02-08)
The transcription factor OCT-4 is regarded as a critical factor in controlling mammalian early embryonic development because of its role in toti-/pluripotency. In human preimplantation embryos, OCT-4 studies are limited to RNA analysis of abnormally developing embryos. This study thoroughly
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F9012-1ML | 04061837517280 |