Skip to Content
Merck

F8517

Flow adapter for Luer lock LC columns

column I.D. 2.5 cm

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
SB.52
UNSPSC Code:
41115716
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

LPLC: suitable

column I.D.

2.5 cm

General description

Flow adapters consist of 1/16 in. O.D. FEP tubing, a polyacetal body, a Viton® O-ring seal and a 20 μm porosity HDPE bed support. One replacement bed support is supplied with each flow adapter. One adapter can be combined with a standard Luer Lock, non-jacketed column to make a column with an adjustable bed volume.

Legal Information

Viton is a registered trademark of E. I. du Pont de Nemours and Company


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library