Skip to Content
Merck
All Photos(1)

Key Documents

F4523

Sigma-Aldrich

Fast Red TR/Naphthol AS-MX

SIGMAFAST, fluorogenic, tablet

Synonym(s):

4-Chloro-2-methylbenzenediazonium / 3-Hydroxy-2-naphthoic acid 2,4-dimethylanilide phosphate tablets

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

Product Name

SIGMAFAST Fast Red TR/Naphthol AS-MX Tablets, tablet, To prepare 10 mL

form

tablet

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Developed for use as a precipitating substrate for the detection of alkaline phosphatase activity in immunochemistry. Primary uses are in immunohistology, immunoblotting, or dot blotting. SIGMAFAST Fast Red TR/Naphthol AS-MX tablets contain 0.6 mM levamisole to block endogenous alkaline phosphatase activity. SIGMAFAST Fast Red TR/Naphthol AS-MX Tablets require no additional steps or buffers to prepare an active substrate solution.
SIGMAFAST Fast Red TR/Naphthol AS-MX tablets has been used in fluorescent in situ hybridization for the analysis of gene expression in neurons and synapses.

Reconstitution

Each tablet set dissolved in 10 mL of deionized water yields a ready-to-use buffered solution.

Legal Information

SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G B Baretton et al.
British journal of cancer, 80(3-4), 546-555 (1999-07-17)
The molecular mechanisms leading to androgen-independent growth in prostate cancer (PC) are poorly understood. Androgen deprivation therapy (ADT) results physiologically in a decrease in proliferation and an increase in programmed cell death (PCD)/apoptosis. The aim of our study was to
Shinjae Chung et al.
Nature, 545(7655), 477-481 (2017-05-18)
In humans and other mammalian species, lesions in the preoptic area of the hypothalamus cause profound sleep impairment, indicating a crucial role of the preoptic area in sleep generation. However, the underlying circuit mechanism remains poorly understood. Electrophysiological recordings and
Ulla R M Holopainen et al.
Journal of the science of food and agriculture, 92(15), 2944-2951 (2012-05-03)
Hordeins are major storage proteins of barley (Hordeum vulgare L.) grains and are considered to influence malting and brewing by forming a matrix surrounding the starch granules which affects the release of fermentable sugars. However, the extent to which environmental
Calcium imaging of basal forebrain activity during innate and learned behaviors
Harrison TC, et al.
Frontiers in Neural Circuits, 10, 36-36 (2016)
The
In Situ Hybridization Methods (2015)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service