Skip to Content
Merck
All Photos(1)

Key Documents

F4425

Sigma-Aldrich

Fostriecin sodium salt from Streptomyces pulveraceus

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26NaO9P
CAS Number:
Molecular Weight:
452.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Streptomyces sp. (Streptomyces pulveraceous)

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

ethanol: soluble
methanol: soluble
water: soluble

storage temp.

−20°C

SMILES string

[Na+].C[C@@](O)(\C=C\[C@H]1CC=CC(=O)O1)[C@@H](C[C@@H](O)\C=C/C=C\C=C\CO)OP(O)([O-])=O

InChI

1S/C19H27O9P.Na/c1-19(23,12-11-16-9-7-10-18(22)27-16)17(28-29(24,25)26)14-15(21)8-5-3-2-4-6-13-20;/h2-8,10-12,15-17,20-21,23H,9,13-14H2,1H3,(H2,24,25,26);/q;+1/p-1/b3-2-,6-4+,8-5-,12-11+;/t15-,16+,17+,19+;/m0./s1

InChI key

XBUIKNRVGYFSHL-IAVQPKKASA-M

Application

Fostriecin has been used as a protein phosphatases type 2A (PP2A) inhibitor to study its effects on phosphorylated myosin light chain (pMLC) levels in axon initial segments (AIS). It has also been used as a PP2A inhibitor to study its effects on the regulation of protein kinase Akt (pAKT) by dopamine (DA) in larval zebrafish brain.

Biochem/physiol Actions

Fostriecin is a natural inhibitor of serine/ threonine phosphatases. It is a phosphate monoester produced by fermentation beer of Streptomyces pulveraceus. Fostriecin is a strong inhibitor of protein phosphatases type 2A and 4 (PP2A and PP4) and a weak inhibitor of protein phosphatases type 1 and 5 (PP1 and PP5). It exhibits anti-tumor activity against several tumor cells in vitro and L1210 and P388 leukemias in vivo. The anti-tumor activity of fostriecin is due to its ability to interfere with the reversible phosphorylation of proteins that are involved in the progression of the cell cycle. It also exhibits cytotoxic effects. Fostriecin can inhibit the DNA, RNA and protein synthesis.

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Serine/Threonine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Packaged under argon.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Stampwala et al.
The Journal of antibiotics, 36(12), 1601-1605 (1983-12-01)
A complex of structurally related compounds that exhibit in vivo antileukemic activity was isolated from fermentation broths of a new streptomycete. The components of this complex are water soluble phosphate esters containing a conjugated triene system. The isolation and characterization
A Cheng et al.
Cancer research, 58(16), 3611-3619 (1998-08-29)
Fostriecin, a structurally unique phosphate ester, is presently under evaluation in clinical trials to determine its potential use as an antitumor drug in humans. Fostriecin has been reported as having inhibitory activity against DNA topoisomerase type II and protein phosphatases
Fostriecin, an antitumor antibiotic with inhibitory activity against serine/threonine protein phosphatases types 1 (PP1) and 2A (PP2A), is highly selective for PP2A
Walsh A, et al.
Febs Letters, 416(3), 230-234 (1997)
C J Hastie et al.
FEBS letters, 431(3), 357-361 (1998-08-26)
Protein phosphatase 4 (PP4) is a protein serine/threonine phosphatase that predominantly localises to centrosomes and plays a role in microtubule organisation at centrosomes. Here, PP4 catalytic subunit has been purified from porcine testis to near homogeneity and a specific activity
A H Walsh et al.
FEBS letters, 416(3), 230-234 (1997-12-31)
Fostriecin, an antitumor antibiotic produced by Streptomyces pulveraceus, is a strong inhibitor of type 2A (PP2A; IC50 3.2 nM) and a weak inhibitor of type 1 (PP1; IC50 131 microM) serine/threonine protein phosphatases. Fostriecin has no apparent effect on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service