Skip to Content
Merck
All Photos(1)

Key Documents

F2878

Sigma-Aldrich

Ficoll® PM 70

Type 70

Synonym(s):

Poly(sucrose-co-epichlorhydrin)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

type

Type 70

Quality Level

form

powder

mol wt

~70,000

color

white

solubility

H2O: soluble 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

Looking for similar products? Visit Product Comparison Guide

General description

Ficoll® PM70, a high molecular weight branched polysaccharide polymer, is believed that they do not contain any ionized groups. This sucrose-polymer is formed by the copolymerization of sucrose with epichlorhydrin.

Application

Ficoll® PM 70 has been used:
  • as a macromolecule in a comparative study to evaluate its effect in vitrification solutions
  • as a supplement in the vitrification solution for the vitrification procedure of oocytes
  • as a component of low serum medium for human lung fibroblast cell culture to promote a crowded environment

Biochem/physiol Actions

Ficoll is commonly used as a macromolecular constituent of vitrification solutions. Due to their dense branching, Ficolls also serve as ideal, nearly spherical molecular crowding agents.
FicollTM PM 70 is used as a macromolecular crowding agent in studies of cell volume signaling and protein refolding.It may be used in tissue engineering and macromolecular conformation research for the development, evaluation, and use of macromolecular crowding (MMC) systems and configurations. Ficoll PM-70 can also be used in vitrification research.

Other Notes

A nonionic synthetic polymer of sucrose.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Checura et al.
Theriogenology, 67(5), 919-930 (2006-12-19)
This study was designed to evaluate vitrification procedures for in vitro matured bovine oocytes for efficient blastocyst production after warming, IVF and culture. A second goal was to replace serum as the macromolecular component of the vitrification solution, without compromising
M B Burg
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 10(5-6), 251-256 (2000-12-23)
The non-ideal properties of solutions containing high concentrations of macromolecules can result in enormous increases in the activity of the individual macromolecules. There is considerable evidence that macromolecular crowding and confinement not only occur in cells, but that these are
Y Georgalis et al.
Journal of colloid and interface science, 386(1), 141-147 (2012-08-23)
Ficoll PM70 is an important branched polysaccharide polymer with a variety of applications in biology and biophysics. The accepted picture for the aqueous solutions of this polymer is that of a polydisperse nearly spherical suspension with a molecular weight of
M Al-Habori
The international journal of biochemistry & cell biology, 33(9), 844-864 (2001-07-20)
Macromolecular crowding has been proposed as a mechanism by means of which a cell can sense relatively small changes in volume or, more accurately, the concentration of intracellular solutes. According to the macromolecular theory, the kinetics and equilibria of enzymes
Emmanuel Dauty et al.
Journal of molecular recognition : JMR, 17(5), 441-447 (2004-09-14)
Aqueous environments in living cells are crowded, with up to >50 wt% small and macromolecule-size solutes. We investigated quantitatively one important consequence of molecular crowding--reduced diffusion of biologically important solutes. Fluorescence correlation spectroscopy (FCS) was used to measure the diffusion

Protocols

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service