Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
mouse
Quality Segment
conjugate
FITC conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
DM1A, monoclonal
form
buffered aqueous solution
mol wt
antigen ~50 kDa
species reactivity
yeast, mouse, amphibian, human, rat, chicken, fungi, bovine
storage condition
protect from light
technique(s)
direct immunofluorescence: 1:50 using cultured BHK cells
isotype
IgG1
UniProt accession no.
application(s)
research pathology
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TUBA4A(7277)
mouse ... Tuba1a(22142)
rat ... Tuba1a(64158)
General description
Monoclonal Anti-α-Tubulin (mouse IgG1 isotype) is derived from the DM1A hybridoma produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice. Purified chick brain microtubules were used as immunogen. The isotype is determined by a double diffusion immunoassay using Mouse Monoclonal Antibody Isotyping Reagents, Product Number ISO2. The product is Protein A purified Monoclonal Anti-α-Tubulin antibody conjugated to fluorescein isothiocyanate, isomer I. It is purified by gel filtration and contains no detectable free FITC.
Immunogen
Application
- respiratory epithelium tissue in a study to determine the tubulin expression in the mice cilia
- breast cancer tissue sections to study the effect of LMO4 on the centrosome amplification and mitotic spindle abnormalities
- spindle and chromosomes of oocytes
- detection of tubulin by immunofluorescence and confocal microscopy in lung carcinoma cell.
- immunofluorescent staining of microtubules in human embryos and mitotic spindles from spleen lymphoblast.
Biochem/physiol Actions
Physical form
Preparation Note
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form purified immunoglobulin | antibody form ascites fluid | antibody form purified from hybridoma cell culture | antibody form purified immunoglobulin |
| conjugate FITC conjugate | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone DM1A, monoclonal | clone DM1A, monoclonal | clone B-5-1-2, monoclonal | clone 6-11B-1, monoclonal |
| Gene Information human ... TUBA4A(7277) | Gene Information human ... TUBA4A(7277) | Gene Information human ... TUBA4A(7277) | Gene Information human ... TUBA4A(7277) |
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| species reactivity yeast, mouse, amphibian, human, rat, chicken, fungi, bovine | species reactivity yeast, mouse, amphibian, human, rat, chicken, fungi, bovine | species reactivity mouse, chicken, Chlamydomonas, African green monkey, human, rat, bovine, sea urchin, kangaroo rat | species reactivity bovine, frog, invertebrates, human, hamster, mouse, protista, pig, monkey, chicken, rat, plant |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
What is the immunogen for Monoclonal Anti-α-Tubulin antibody, clone DM1A?
1 answer-
The immunogen for Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A is microtubules from chicken embryo brain. This antibody recognizes an epitope located at the C-terminal end of the α-tubulin isoform (amino acids 426-430) as noted in the following references:Breitling, F., and Little, M., J. Molec. Biol., 189, 367-370 (1986).Wolff, A., et al., Biol. Cell, 63, 319-326 (1988).Serrano, L., et al., Anal. Biochem., 159, 253-259 (1986).
Helpful?
-



