Skip to Content
Merck
All Photos(1)

Key Documents

F1638

Sigma-Aldrich

Anti-Pig IgG (whole molecule)−FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Porcine IgG (whole molecule)−FITC antibody produced in rabbit, Anti-Swine IgG (whole molecule)−FITC antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:32

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates various cellular immunological responses. Pig IgGs are involved in mediating several pathogenic responses, including the response to Trichinella infections . Anti-Pig IgG (whole molecule)−FITC antibodies can be used to study the detection and localization of pig IgGs. The product is specific for pig IgGs.
Pig IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in pig serum.

Specificity

Rabbit polyclonal anti-Pig IgG (whole molecule)?FITC antibody reacts with pig IgG in vitro, in pig serum and biological fluids.

Immunogen

Purified pig IgG

Application

Anti-Pig IgG (whole molecule)-FITC antibody is suitable for use in direct immunofluorescence (1:32).
Anti-Pig IgG (whole molecule)-FITC antibody produced in rabbit has been used in indirect immunofluorescence assay.
Rabbit polyclonal anti-Pig IgG (whole molecule)−FITC antibody may be used to detect and quantitate the level of IgG in pig serum and biological fluids by fluorescent techniques. It may also be used as a secondary antibody in assays that use pig IgG as the primary antibody.

Biochem/physiol Actions

IgG antibody is the most abundant serum antibody among immunoglobulins of the immune system. It is found in blood and in extracellular fluid when exposed to an antigen. It protects from bacterial, fungal and viral infections. Maternal IgG crosses from mother to fetus through the placental barrier by pinocytosis to provide passive immunity for the neonate and protect against infections.
IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection. Fluorescein isothiocyanate (FITC) is a fluorescein derivative (fluorochrome) used to tag antibodies, including secondary antibodies, for use in fluorescence-based assays and procedures. FITC excites at 495 nm and emits at 521 nm.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for a maximum of one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chunyun Qi et al.
iScience, 25(10), 105268-105268 (2022-10-25)
Classical swine fever virus (CSFV), a classic swine fever pathogen, causes severe economic losses worldwide. Poly (rC)-binding protein 1 (PCBP1), which interacts with Npro of CSFV, plays a vital role in CSFV growth. We are the first to report the
Guanylate-binding protein 1, an interferon-induced GTPase, exerts an antiviral activity against classical swine fever virus depending on its GTPase activity
Li LF, et al.
Journal of virology, 90(9), 4412-4426 (2016)
Placental structure, function and drug transfer
Griffiths SK and Campbell JP
Encyclopedia of Chemical Processing and Design, 15(2), 84-89 (2014)
Changchao Huan et al.
Viruses, 14(8) (2022-08-27)
Pseudorabies virus (PRV) is one of the most important pathogens causing serious diseases and leads to huge economic losses in the global swine industry. With the continuous emergence of PRV variants and the increasing number of cases of human infection
Zicong Xie et al.
PLoS pathogens, 14(12), e1007193-e1007193 (2018-12-14)
Classical swine fever (CSF) caused by classical swine fever virus (CSFV) is one of the most detrimental diseases, and leads to significant economic losses in the swine industry. Despite efforts by many government authorities to stamp out the disease from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service