Skip to Content
Merck
All Photos(1)

Documents

F1293

Sigma-Aldrich

Finasteride

≥98% (HPLC), powder

Synonym(s):

N-(2-methyl-2-propyl)-3-oxo-4-aza-5α-androst-1-ene-17β-carboxamide, N-tert-Butyl-3-oxo-4-aza-5α-androst-1-en-17β-carboxamide, MK-906

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36N2O2
CAS Number:
Molecular Weight:
372.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: soluble >20 mg/mL

storage temp.

room temp

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(=O)NC(C)(C)C)[C@@]4(C)CC[C@]3([H])[C@@]1(C)C=CC(=O)N2

InChI

1S/C23H36N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h11,13-18H,6-10,12H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1

InChI key

DBEPLOCGEIEOCV-WSBQPABSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Finasteride has been used:
  • as a component of L-15 media to investigate the effects of finasteride exposure on brain, liver and testis tissues
  • in culture medium to treat tumor cells in vitro
  • as a positive control to treat benign prostatic hyperplasia (BPH)
  • to examine its effect on structural and molecular biology on the ventral prostate of senile mice

Biochem/physiol Actions

Finasteride can reduce serum and scalp dihydrotestosterone (DHT) by preventing the transformation of testosterone to DHT. It is usually used to manage benign prostate hyperplasia and pattern hair loss in male. Finasteride possesses androgen-distracting properties.
Selective 5α-reductase inhibitor; antiandrogen.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Joseph et al.
Experimental physiology, 103(2), 276-290 (2017-11-24)
What is the central question of this study? What are the contributions of allopregnanolone, the neuroactive metabolite of progesterone, and nuclear (nPR) and membrane (mPR) progesterone receptors to the respiratory effect of progesterone in newborn rats? What is the main
Finasteride adverse effects: An update
Carreno-Orellana N, et al.
Revista Brasileira de Farmacognosia, 144(12), 1584-1590 (2016)
Douglas J Fort et al.
Journal of applied toxicology : JAT, 39(7), 1066-1078 (2019-03-09)
The impact of the perfluoro-chemical, perfluorooctanesulfonate (PFOS), on gonadal steroidogenesis during sexual differentiation in Silurana tropicalis was examined because of its ubiquity in the environment, bioaccumulative nature and potential to disturb endocrine activity. A partial life cycle study exposing S.
Yu-Liang Miao et al.
PloS one, 9(7), e101450-e101450 (2014-07-06)
Post-traumatic stress disorder (PTSD) is a debilitating anxiety disorder that may develop after an individual has experienced or witnessed a severe traumatic event. It has been shown that the 18 kDa translocator protein (TSPO) may be correlated with PTSD and
Inhibitory effects of Ponciri Fructus on testosterone-induced benign prostatic hyperplasia in rats
Jeon W Y, et al.
BMC Complementary and Alternative Medicine, 17(1), 384-384 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service